Skip to Content
Merck
All Photos(2)

Key Documents

323527

Sigma-Aldrich

Selenium tetrachloride

Synonym(s):

Selenium (Ⅳ) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SeCl4
CAS Number:
Molecular Weight:
220.77
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder or crystals

grade

for analytical purposes

form

powder or crystals

concentration

35.0-36.5% Se (after reduction with sulphite, gravimetric)

density

2.6 g/mL at 25 °C (lit.)

SMILES string

Cl[Se](Cl)(Cl)Cl

InChI

1S/Cl4Se/c1-5(2,3)4

InChI key

LNBXMNQCXXEHFT-UHFFFAOYSA-N

Related Categories

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sun-Chol Yu et al.
Organic & biomolecular chemistry, 8(4), 828-834 (2010-02-06)
Low molecular weight seleno-organic compounds exhibit glutathione peroxidase (GPx)-like activity; the well-known compound ebselen is being used in clinical trials as a stroke medication. Here, we describe the facile one-step synthesis of novel 5-selenized salicylic acid derivatives using selenium tetrachloride.
M K Christensen et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 40(4), 575-579 (1992-04-01)
The autometallographic retrograde tracing of zinc-containing neurons by intracerebral injection of sodium selenite (Na2SeO3), introduced by Danscher in 1982, has recently been described in more detail. Intracerebral injections of both sodium selenide (Na2Se) and sodium selenite (Na2SeO3) have been successfully
Abdul Majeed et al.
Journal of materials chemistry. B, 8(36), 8444-8458 (2020-08-20)
A facile, green synthesis of selenium doped zinc oxide nano-antibiotic (Se-ZnO-NAB) using the Curcuma longa extract is reported to combat the increased emergence of methicillin-resistant Staphylococcus aureus (MRSA). The developed Se-ZnO-NAB were characterized for their physicochemical parameters and extensively evaluated
F Islam et al.
Archives of biochemistry and biophysics, 289(1), 161-166 (1991-08-15)
Various inorganic selenocompounds dose-dependently inhibited the rat brain prostaglandin (PG) D synthase, both in the purified enzyme preparation and in the crude brain supernatant. All of the quadrivalent selenium compounds tested had a very limited range of IC50 values in
R Takahata et al.
Brain research, 623(1), 65-71 (1993-09-24)
Prostaglandin (PG) D2 has been postulated to be an endogenous sleep-promoting factor. Biosynthesis of PGD2 is catalyzed by PGD synthase (prostaglandin-H2 D-isomerase, EC 5.3.99.2), the activity of which is inhibited by inorganic selenium compounds such as SeCl4 and Na2SeO3. We

Articles

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service