Skip to Content
Merck
All Photos(1)

Key Documents

230499

Sigma-Aldrich

Selenourea

98%

Synonym(s):

2-Selenourea, Carbamimidoselenoic acid, Isoselenourea, Selenouronium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CSeNH2
CAS Number:
Molecular Weight:
123.02
Beilstein:
1734744
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

mp

210-215 °C (dec.)

functional group

amine

SMILES string

NC(N)=[Se]

InChI

1S/CH4N2Se/c2-1(3)4/h(H4,2,3,4)

InChI key

IYKVLICPFCEZOF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Selenourea [(SeC(NH2)2)] has been used as a selenium (Se) precursor for the insertion of Se into protein crystals.
It can also be used as a reagent to synthesize:
  • Selenazoles from α -bromo ketones presence of β -cyclodextrin.
  • Symmetrical diaryl selenides by C-Se cross coupling reaction in presence of coper oxide nanaoparticles as a catalyst.
  • CdSe nanocrystals.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zakhele Ndala et al.
Nanomaterials (Basel, Switzerland), 10(9) (2020-09-13)
Herein we report on the use of different metal precursors in the synthesis of MoSe2 nanomaterials in order to control their morphology. The use of Mo(CO)6 as the metal precursor resulted in the formation of wrinkled few-layer nanosheets, while the
A novel synthesis of CdSe nanocrystals
Epifani M, et al.
Materials Letters, 58(19), 2429-2432 (2004)
Sung Min Kwon et al.
Science advances, 4(4), eaap9104-eaap9104 (2018-04-18)
We report a general strategy for obtaining high-quality, large-area metal-chalcogenide semiconductor films from precursors combining chelated metal salts with chalcoureas or chalcoamides. Using conventional organic solvents, such precursors enable the expeditious formation of chalco-gels, which are easily transformed into the
B Mishra et al.
The journal of physical chemistry. A, 110(5), 1894-1900 (2006-02-03)
Pulse radiolysis coupled with absorption detection has been employed to study one-electron oxidation of selenomethionine (SeM), selenocystine (SeCys), methyl selenocysteine (MeSeCys), and selenourea (SeU) in aqueous solutions. Hydroxyl radicals (*OH) in the pH range from 1 to 7 and specific
Cristina Trujillo et al.
The journal of physical chemistry. B, 112(17), 5479-5486 (2008-04-09)
The gas-phase reactions between Ca(2+) and selenourea were investigated by means of electrospray/tandem mass spectrometry techniques. The MS/MS spectra of [Ca(selenourea)](2+) complexes show intense peaks at m/z 43, 121, 124, and 146 and assigned to monocations produced in different coulomb

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service