Skip to Content
Merck
All Photos(1)

Documents

310824

Sigma-Aldrich

D-Glucose-2-d

98 atom % D, 99% (CP)

Synonym(s):

Deuterated Glucose, Labeled Glucose, Dextrose-2-d

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2[CHOH]3CDOHCHO
CAS Number:
Molecular Weight:
181.16
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

99% (CP)

form

powder

optical activity

[α]25/D +52.0°, c = 2 in H2O (trace NH4OH)

mp

150-152 °C (lit.)

mass shift

M+1

SMILES string

[2H][C@](O)(C=O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h1,3-6,8-12H,2H2/t3-,4+,5+,6+/m0/s1/i3D

InChI key

GZCGUPFRVQAUEE-FPCZXLAFSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cecile Vernochet et al.
Molecular and cellular biology, 29(17), 4714-4728 (2009-07-01)
White adipose tissue (WAT) stores energy in the form of triglycerides, whereas brown tissue (BAT) expends energy, primarily by oxidizing lipids. WAT also secretes many cytokines and acute-phase proteins that contribute to insulin resistance in obese subjects. In this study
Jill Pecon-Slattery et al.
BMC genomics, 9, 66-66 (2008-02-07)
Feline immunodeficiency virus (FIV) naturally infects multiple species of cat and is related to human immunodeficiency virus in humans. FIV infection causes AIDS-like disease and mortality in the domestic cat (Felis catus) and serves as a natural model for HIV
Hyun Woo Jeong et al.
Diabetes, 60(2), 496-506 (2011-01-29)
Peroxisome proliferator-activated receptor (PPAR)-α/γ dual agonists have been developed to alleviate metabolic disorders. However, several PPARα/γ dual agonists are accompanied with unwanted side effects, including body weight gain, edema, and tissue failure. This study investigated the effects of a novel
Guillaume Andre et al.
Journal of bacteriology, 190(21), 7079-7086 (2008-09-02)
The lysin motif (LysM) is a ubiquitous protein module that binds peptidoglycan and structurally related molecules. Here, we used single-molecule force spectroscopy (SMFS) to measure and localize individual LysM-peptidoglycan interactions on both model and cellular surfaces. LysM modules of the
Sharareh Siamakpour-Reihani et al.
PloS one, 6(6), e20412-e20412 (2011-06-16)
Tacrolimus (FK506) is an immunosuppressive drug that binds to the immunophilin FKBPB12. The FK506-FKBP12 complex associates with calcineurin and inhibits its phosphatase activity, resulting in inhibition of nuclear translocation of nuclear factor of activated T-cells (NFAT). There is increasing data

Articles

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service