Skip to Content
Merck
All Photos(1)

Key Documents

294381

Sigma-Aldrich

N,N′-(1,2-Dihydroxyethylene)bisacrylamide

97%

Synonym(s):

N,N′-Bisacryloyl-1,2-dihydroxy-1,2-ethylenediamine, DHEBA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[H2C=CHCONHCH(OH)-]2
CAS Number:
Molecular Weight:
200.19
Beilstein:
1941875
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

mp

156 °C (dec.) (lit.)

SMILES string

OC(NC(=O)C=C)C(O)NC(=O)C=C

InChI

1S/C8H12N2O4/c1-3-5(11)9-7(13)8(14)10-6(12)4-2/h3-4,7-8,13-14H,1-2H2,(H,9,11)(H,10,12)

InChI key

ZMLXKXHICXTSDM-UHFFFAOYSA-N

General description

N,N′-(1,2-Dihydroxyethylene)bisacrylamide, commonly referred to as DHEBA or bis-acrylamide. It is used as a crosslinking agent in various biomedical applications. Its structure includes two acrylamide groups linked by a dihydroxyethylene bridge, which enhances its reactivity and ability to form hydrogels. In the biomedical field, DHEBA is primarily utilized in the synthesis of hydrogels that can be used for drug delivery systems, tissue engineering scaffolds, and biosensors. The hydrophilic nature of the compound allows for the formation of hydrogels that can retain water and biological molecules, making them suitable for applications that require biocompatibility and bioactivity.

Application

N,N′-(1,2-Dihydroxyethylene)bisacrylamide can be used as:
  • A crosslinker in the synthesis of hydrogel microparticles with a porous hydrogel structure. This porosity is essential for enhancing biomolecule accessibility and mass transport, which are critical for the performance of biosensing applications.
  • A cross-linking agent in the preparation of molecularly imprinted polymers, that can selectively bind to oxidized low-density lipoprotein (oxLDL). The cross-linking enhances the mechanical stability and structural integrity of the polymer matrix.
  • A cross-linking agent in the preparation of hydrogels or polymer matrices that support biological imaging applications.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Suticha Chunta et al.
Talanta, 221, 121549-121549 (2020-10-21)
Very-low-density lipoprotein (VLDL) contributes to the buildup of atherosclerotic plaque in the arteries and can lead to coronary heart disease. In clinical laboratory testing, the cholesterol content of VLDL (VLDL-C) cannot be assessed directly by the enzymatic colorimetric assay as
Suticha Chunta et al.
Analytica chimica acta, 1116, 27-35 (2020-05-12)
Oxidized-low-density lipoprotein (oxLDL) is well-recognized as an actual patho-atherogenic lipoprotein: elevated serum concentration of oxLDL increases the risk for developing atherosclerosis, leading to coronary artery disease (CAD). Herein, we report an approach for sensing oxLDL directly in serum with molecularly
Gisela Giorgi et al.
Experimental physiology, 100(10), 1199-1216 (2015-06-06)
What is the central question of this study? The aim was to explore the role and hitherto unclear mechanisms of action of iron proteins in protecting the lung against the harmful effects of iron accumulation and the ability of pulmonary
U Neumann et al.
Analytical biochemistry, 206(1), 1-5 (1992-10-01)
A simple and rapid method for the quantitation of proteins separated either by sodium dodecyl sulfate-electrophoresis or by isoelectric focusing in slab gels is presented. The method is based on the solubility of polyacrylamide gels crosslinked with N, N'-1, 2-dihydroxyethylenebisacrylamide
Krongkaew Navakul et al.
Nanomedicine : nanotechnology, biology, and medicine, 13(2), 549-557 (2016-08-26)
Dengue fever is a major disease that kills many people in the developing world every year. During early infection, a patient displays a high temperature without other signs. After this stage, and without proper treatment, serious damage to internal organs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service