Skip to Content
Merck
All Photos(1)

Documents

293741

Sigma-Aldrich

Succinic anhydride-2,2,3,3-d4

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4D4O3
CAS Number:
Molecular Weight:
104.10
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Assay

99% (CP)

form

solid

technique(s)

protein expression: suitable

bp

261 °C

mp

118-120 °C (lit.)

mass shift

M+4

SMILES string

[2H]C1([2H])C(=O)OC(=O)C1([2H])[2H]

InChI

1S/C4H4O3/c5-3-1-2-4(6)7-3/h1-2H2/i1D2,2D2

InChI key

RINCXYDBBGOEEQ-LNLMKGTHSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service