Skip to Content
Merck
All Photos(1)

Key Documents

292753

Sigma-Aldrich

Pentaethylenehexamine

technical grade

Synonym(s):

3,6,9,12-Tetraazatetradecane-1,14-diamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2NH(CH2CH2NH)3CH2CH2NH2
CAS Number:
Molecular Weight:
232.37
Beilstein:
1768042
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

refractive index

n20/D 1.5096 (lit.)

solubility

water: soluble 500 g/L at 20 °C

density

0.95 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

NCCNCCNCCNCCNCCN

InChI

1S/C10H28N6/c11-1-3-13-5-7-15-9-10-16-8-6-14-4-2-12/h13-16H,1-12H2

InChI key

LSHROXHEILXKHM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pentaethylenehexamine (PEHA) is an acyclic polyamine that can be used to prepare amine-modified adsorbent.

Application

5.Pentaethylenehexamine has been used in the preparation of:
  • polyisobutylene succinimide (PIBSI) dispersants, used as engine oil-additives
  • poly(glycidyl methacrylate) (P(GMA)) homopolymers
  • poly(amidoamine)s

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solmaz Pirouz et al.
The journal of physical chemistry. B, 118(14), 3899-3911 (2014-03-19)
A novel methodology based on fluorescence quenching measurements is introduced to determine quantitatively the amine content of polyisobutylene succinimide (PIBSI) dispersants used as engine oil-additives. To this end, a series of five PIBSI dispersants were prepared by reacting 2 mol
Hua Wei et al.
Biomacromolecules, 14(1), 275-284 (2012-12-18)
Polyethylenimine (PEI) is one of the most broadly used polycations for gene delivery due to its high transfection efficiency and commercial availability but materials are cytotoxic and often polydisperse. The goal of current work is to develop an alternative family
Zhe Xu et al.
International journal of biological macromolecules, 130, 117-124 (2019-02-27)
Nitrilase-catalyzed regioselective hydrolysis of 1-cyanocyclohexaneacetonitrile (1-CHAN) is a green and efficient approach for the preparation of 1-cyanocyclohexaneacetic acid (1-CHAA), a key precursor for the synthesis of gabapentin. Here, a mesoporous biosilica particles prepared by the ethyleneamine-mediated silicification have been used
Hong Li et al.
Journal of drug targeting, 26(5-6), 466-473 (2018-01-30)
RX-0201 is an antisense oligonucleotide (ASO) against Akt1 currently in clinical trial for metastatic renal cancer. To improve the delivery of RX-0201 using folate receptor-targeted lipid-albumin nanoparticles (F-LAN). F-LAN were synthesized with the composition of DOTAP/soyPC/TPGS/folate-PEG-DSPE (25:70:4:1 m/m), a cationic human
Yan Ping et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 203, 139-146 (2018-06-05)
Pentaethylenehexamine and d-penicillamine co-functionalized graphene quantum dots (PEHA-GQD-DPA) was made via one two-step thermal pyrolysis. The resulting PEHA-GQD-DPA is composed of the graphene sheets with an average size of 3.16 nm and the rich of functional groups. It gives an ultra

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service