Skip to Content
Merck
All Photos(3)

Key Documents

292265

Sigma-Aldrich

N,N′-Diphenyl-p-phenylenediamine

98%

Synonym(s):

N,N′-Diphenyl-1,4-phenylenediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHC6H4NHC6H5
CAS Number:
Molecular Weight:
260.33
Beilstein:
2215944
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

bp

220-225 °C/0.5 mmHg (lit.)

mp

143-145 °C (lit.)

SMILES string

N(c1ccccc1)c2ccc(Nc3ccccc3)cc2

InChI

1S/C18H16N2/c1-3-7-15(8-4-1)19-17-11-13-18(14-12-17)20-16-9-5-2-6-10-16/h1-14,19-20H

InChI key

UTGQNNCQYDRXCH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • TPA-induced inhibition of gap junctional intercellular communication is not mediated through free radicals.: This research explores the role of N,N′-Diphenyl-p-phenylenediamine (DPPD) in cellular communication, providing insights into its applications in high-performance rubber stabilizers and life science manufacturing chemicals. (Hasler et al., 1990).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Campos-Neto et al.
Tuberculosis (Edinburgh, Scotland), 81(5-6), 353-358 (2002-01-22)
Although the tuberculin test has aided in the diagnosis of tuberculosis for more than 85 years, its interpretation is difficult particularly because sensitization with non-tuberculous mycobacteria leads to false positive tests. Using the guinea pig model of tuberculosis, we have
I Rath et al.
Zeitschrift fur Naturforschung. C, Journal of biosciences, 55(11-12), 957-964 (2001-02-24)
The primary reactions leading to Al toxicity in plant cells have not yet been elucidated. We used soybean (Glycine max [L.] Merr.) cell suspension cultures to address the question whether lipid peroxidation plays an important role in Al toxicity. Upon
Hirokazu Doi et al.
Chemico-biological interactions, 183(3), 363-368 (2009-12-02)
Salicylic acid is a widely used nonsteroidal anti-inflammatory drug (NSAID). But it is known to cause serious liver damage occasionally. Mitochondrial dysfunction and oxidative stress are predicted to be the major factors of salicylic acid-induced liver injury. We investigated the
J R Bishop et al.
Molecular and biochemical parasitology, 118(1), 33-40 (2001-11-13)
It has been known for almost a century that normal human serum can lyse the extracellular blood parasite Trypanosoma brucei brucei. This process is a result of a non-immune killing factor in human sera known as trypanosome lytic factor (TLF).
Hyonseok Hwang
The journal of physical chemistry. A, 113(16), 4780-4787 (2008-11-28)
Coarse-grained (CG) molecular dynamics (MD) simulations are performed to study the insertion of cyclic peptide nanotubes into cell membranes and to examine whether cyclic peptide nanotubes can function as an ion channel and thereby as an antibacterial agent. To do

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service