Skip to Content
Merck
All Photos(5)

Key Documents

283746

Sigma-Aldrich

4-(Aminomethyl)benzoic acid

97%, for peptide synthesis

Synonym(s):

PAMBA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCH2C6H4CO2H
CAS Number:
Molecular Weight:
151.16
Beilstein:
1100606
EC Number:
MDL number:
UNSPSC Code:
12352106
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Product Name

4-(Aminomethyl)benzoic acid, 97%

Quality Level

Assay

97%

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

mp

≥300 °C (lit.)

application(s)

peptide synthesis

SMILES string

NCc1ccc(cc1)C(O)=O

InChI

1S/C8H9NO2/c9-5-6-1-3-7(4-2-6)8(10)11/h1-4H,5,9H2,(H,10,11)

InChI key

QCTBMLYLENLHLA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-(Aminomethyl)benzoic acid (PAMBA) can be used in the synthesis of:
  • Cobalt carboxy phosphonates.
  • Apoptozole (Az), which has cellular potency to promote membrane trafficking of mutant cystic fibrosis transmembrane conductance regulator (CFTR) chloride channel activity.
  • Cyclopeptide composed of L-glutamic acid and 3-aminobenzoic acid that can be used as receptors for a variety of cations and anions.
  • A bioactive peptide which has potent GPR54 (a G protein-coupled receptor) agonistic activity.

Other Notes

4-(Aminomethyl)benzoic acid is also an antihemorrhagic agent, which is used for the treatment of internal haemorrhage.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M N Iakushenko et al.
Zhurnal mikrobiologii, epidemiologii, i immunobiologii, (1)(1), 77-79 (1998-04-09)
The examination of 49 newborn infants revealed that at the early neonatal period the character of the microbial colonization of the intestine depended on the kind of perinatal pathology: in lesions of the central nervous system and conjugation jaundice the
G A Chalyĭ et al.
Zhurnal mikrobiologii, epidemiologii, i immunobiologii, (1)(1), 62-65 (1993-01-01)
The influence of protease-inhibiting preparations on the development of humoral immune response in diseases involving the development of secondary immunodeficiency (experimentally induced acute pancreatitis and staphylococcal infection) has been studied. Five injections of contrycal and epsilon-aminocaproic acid (epsilon-ACA), starting from
Sandra Osburn et al.
Rapid communications in mass spectrometry : RCM, 21(21), 3409-3419 (2007-09-29)
Formation of [bn+17+cat]+ is a prominent collision-induced dissociation (CID) pathway for Li+- and Na+-cationized peptides. Dissociation of protonated and Ag+-cationized peptides instead favors formation of the rival bn+/[bn-1+cat]+ species. In this study the influence of a 4-aminomethylbenzoic acid (4AMBz) residue
A small molecule that binds to an ATPase domain of Hsc70 promotes membrane trafficking of mutant cystic fibrosis transmembrane conductance regulator.
Cho H J, et al.
Journal of the American Chemical Society, 133(50), 20267-20276 (2011)
Carlos F R A C Lima et al.
The journal of physical chemistry. A, 111(42), 10598-10603 (2007-10-05)
The standard (p0 = 0.1 MPa) molar enthalpies of combustion of six aminomethylbenzoic acids were measured at T = 298.15 K by static bomb calorimetry. With these values, the standard molar enthalpies of formation in the crystalline state were obtained.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service