Skip to Content
Merck
All Photos(3)

Key Documents

119695

Sigma-Aldrich

4-(Methylamino)benzoic acid

97%

Synonym(s):

4-(N-Methylamino)benzoic acid, N-Methyl-4-aminobenzoic acid, p-Carboxy-N-methylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3NHC6H4CO2H
CAS Number:
Molecular Weight:
151.16
Beilstein:
2082805
EC Number:
MDL number:
UNSPSC Code:
12352106
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

reaction suitability

reaction type: solution phase peptide synthesis

mp

160-162 °C (lit.)

application(s)

peptide synthesis

SMILES string

CNc1ccc(cc1)C(O)=O

InChI

1S/C8H9NO2/c1-9-7-4-2-6(3-5-7)8(10)11/h2-5,9H,1H3,(H,10,11)

InChI key

ZVIDMSBTYRSMAR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea Martorana et al.
Molecular bioSystems, 7(11), 2967-2969 (2011-09-14)
Spectroscopic measurements combined with Density Functional Theory calculations were applied to the characterization of the homocoupling reaction of 4-methylamino benzoic acid mediated by laccase.
Shoujiro Ogawa et al.
Journal of pharmaceutical and biomedical analysis, 136, 126-133 (2017-01-13)
The plasma/serum concentration of 25-hydroxyvitamin D
Mitchell P Thayer et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 84(1), 227-232 (2011-10-14)
The local environment dictates the structural and functional properties of many important chemical and biological systems. The impact of pH on the photophysical properties of a series of para-aminobenzoic acids is examined using a combination of experimental spectroscopy and quantum
Farook Adam et al.
Journal of colloid and interface science, 311(1), 135-143 (2007-03-30)
Iron and 4-(methylamino)benzoic acid have been successfully incorporated into silica extracted from rice husk. The silica/Fe/amine complex, RH-Fe(5% amine), showed a ca. 24% increase in specific surface area compared to RH-Fe. This increase was attributed to the templated formation of
Bernd Moritz et al.
Electrophoresis, 38(24), 3136-3146 (2017-09-10)
CZE is a well-established technique for charge heterogeneity testing of biopharmaceuticals. It is based on the differences between the ratios of net charge and hydrodynamic radius. In an extensive intercompany study, it was recently shown that CZE is very robust

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service