Skip to Content
Merck
All Photos(1)

Key Documents

282413

Sigma-Aldrich

2,3-Dimercapto-1-propanol tributyrate

97%

Synonym(s):

BAL tributyrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CO2CH2CH(SCOCH2CH2CH3)CH2SCOCH2CH2CH3
CAS Number:
Molecular Weight:
334.49
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.495 (lit.)

bp

156-160 °C/0.15 mmHg (lit.)

density

1.083 g/mL at 25 °C (lit.)

functional group

ester
thioester

SMILES string

CCCC(=O)OCC(CSC(=O)CCC)SC(=O)CCC

InChI

1S/C15H26O4S2/c1-4-7-13(16)19-10-12(21-15(18)9-6-3)11-20-14(17)8-5-2/h12H,4-11H2,1-3H3

InChI key

NHBKXEKEPDILRR-UHFFFAOYSA-N

Application

2,3-Dimercapto-1-propanol tributyrate is a thioester analog of triacylglycerol and has been used:
  • in lipase inhibition assay during in vitro screening of medicinal plants as antidiabetics with glucosidase and lipase inhibitory activities
  • in colorimetric microplate assay for lipase activity
  • to analyze positional specificity of LipG (lipase-encoding gene) toward triacylglycerol by a simple continuous spectrophotometric method

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Lombard et al.
The Biochemical journal, 358(Pt 3), 773-781 (2001-09-06)
The present study was carried out with a view of determining ricin lipolytic activity on neutral lipids in emulsion and in a membrane-like model. Using 2,3-dimercapto-1-propanol tributyrate (BAL-TC(4)) as substrate, the lipolytic activity of ricin was found to be proportional
[A new simplified colorimetric assay of colipase in duodenal juice using BALB as substrate and its clinical application].
Y Furukawa et al.
Nihon Shokakibyo Gakkai zasshi = The Japanese journal of gastro-enterology, 83(7), 1367-1375 (1986-07-01)
Mi-Hwa Lee et al.
Applied and environmental microbiology, 72(11), 7406-7409 (2006-09-05)
We cloned lipG, which encoded a lipolytic enzyme, from a Korean tidal flat metagenomic library. LipG was related to six putative lipases previously identified only in bacterial genome sequences. These enzymes comprise a new family. We partially characterized LipG, providing
Suk-Jung Choi et al.
Journal of biochemistry and molecular biology, 36(4), 417-420 (2003-08-05)
The present work describes a colorimetric microplate assay for lipase activity based on the reaction between 5,5'-dithiobis(2-nitro benzoic acid) (DTNB) and the hydrolysis product of 2,3-dimercapto-1-propanol tributyrate (DMPTB). Reaction mixtures containing DTNB, DMPTB, and lipase were prepared in microplate wells
Guillermo Ramírez et al.
Evidence-based complementary and alternative medicine : eCAM, 2012, 701261-701261 (2012-10-20)
This work shows the inhibitory effect on glucosidase and lipase enzymes of 23 medicinal plants described as traditional treatments for diabetes in several Mexican sources. Hydroalcoholic extracts of selected plants were evaluated at 1 mg/mL for glucosidase and 0.25 mg/mL for lipase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service