Skip to Content
Merck
All Photos(1)

Key Documents

136948

Sigma-Aldrich

5-Nonanone

98%

Synonym(s):

Dibutyl ketone, Valerone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CO(CH2)3CH3
CAS Number:
Molecular Weight:
142.24
Beilstein:
1743583
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.419 (lit.)

bp

186-187 °C (lit.)

mp

−50 °C (lit.)

density

0.826 g/mL at 25 °C (lit.)

SMILES string

CCCCC(=O)CCCC

InChI

1S/C9H18O/c1-3-5-7-9(10)8-6-4-2/h3-8H2,1-2H3

InChI key

WSGCRAOTEDLMFQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

5-Nonanone can be used as a reactant to prepare:
  • 5-nonanketoxime by reacting with hydroxylamine in water.
  • 4-Nitro-2,6-dipropylphenol by treating with a solution of formyl nitroenamine in the presence of a base.

Biochem/physiol Actions

5-Nonanone induces clinical neuropathy in rats.

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

141.8 °F - closed cup

Flash Point(C)

61 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A convenient method for synthesizing modified 4-nitrophenols
Nakaike Y, et al.
The Journal of Organic Chemistry, 70(24), 10169-10171 (2005)
Commercial-grade methyl heptyl ketone (5-methyl-2-octanone) neurotoxicity: contribution of 5-nonanone.
J L O'Donoghue et al.
Toxicology and applied pharmacology, 62(2), 307-316 (1982-02-01)
Possible role of metabolism in 5-nonanone neurotoxicity.
G D DiVincenzo et al.
Neurotoxicology, 3(1), 55-63 (1982-07-01)
Controlling Selectivity in the Consecutive Reaction Network of Aldoxime Hydrogenation to Primary Amines.
Gebauer-Henke E, et al.
Catalysis Science & Technology, 2, 2539-2548 (2012)
Development of a general Pd (II)-catalyzed intermolecular hydroalkoxylation reaction of vinylphenols by using a sacrificial alcohol as the hydride source
Zhang Y and Sigman MS
Organic Letters, 8(24), 5557-5560 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service