Skip to Content
Merck
All Photos(1)

Documents

277487

Sigma-Aldrich

2-Methyltetrahydro-3-furanone

98%

Synonym(s):

4,5-Dihydro-2-methyl-3(2H)-furanone, 2-Methyltetrahydro-3-furanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8O2
CAS Number:
Molecular Weight:
100.12
Beilstein:
1341334
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.429 (lit.)

bp

139 °C (lit.)

density

1.034 g/mL at 25 °C (lit.)

SMILES string

CC1OCCC1=O

InChI

1S/C5H8O2/c1-4-5(6)2-3-7-4/h4H,2-3H2,1H3

InChI key

FCWYQRVIQDNGBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Methyltetrahydro-3-furanone has been employed as an quorum sensing inhibitor-molecule to investigate its effect on growth and biofilm formation in Hafnia alvei.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

102.2 °F - closed cup

Flash Point(C)

39 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eliseth Souza Viana et al.
Biological research, 42(4), 427-436 (2010-02-09)
The objective of this study was to detect the presence of acyl homoserine lactones (AHLs), signal molecules of the quorum sensing system in biofilm formed by Hafnia alvei strains. It also evaluated the effect of synthetic quorum sensing inhibitors in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service