Skip to Content
Merck
All Photos(1)

Key Documents

W315303

Sigma-Aldrich

5-Ethyl-3-hydroxy-4-methyl-2(5H)-furanone

97%, FG

Synonym(s):

Ethyl fenugreek lactone, Maple furanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10O3
CAS Number:
Molecular Weight:
142.15
FEMA Number:
3153
EC Number:
Council of Europe no.:
2300
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
10.023
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

97%

refractive index

n20/D 1.49 (lit.)

bp

83-86 °C/0.5 mmHg (lit.)

mp

31-35 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

caramel; maple; fruity; sweet

storage temp.

2-8°C

SMILES string

CCC1OC(=O)C(O)=C1C

InChI

1S/C7H10O3/c1-3-5-4(2)6(8)7(9)10-5/h5,8H,3H2,1-2H3

InChI key

IUFQZPBIRYFPFD-UHFFFAOYSA-N

General description

5-Ethyl-3-hydroxy-4-methyl-2(5H)-furanone is one of the key contributors to the aroma of roasted coffee. It occurs naturally in fruits such as blackberry, raspberry and blueberry.

Application


  • RIFM fragrance ingredient safety assessment, 5-ethyl-3-hydroxy-4-methyl-2(5H)-furanone, CAS Registry Number 698-10-2.: This study evaluates the safety of 5-ethyl-3-hydroxy-4-methyl-2(5H)-furanone as a fragrance ingredient, providing comprehensive toxicological data and safety profiles (Api et al., 2024).

  • Elucidating the Odor-Active Aroma Compounds in Alcohol-Free Beer and Their Contribution to the Worty Flavor.: This research identifies and quantifies the aroma compounds responsible for the flavor of alcohol-free beer, highlighting the significant role of 5-ethyl-3-hydroxy-4-methyl-2(5H)-furanone (Piornos et al., 2020).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ali Douaki et al.
Nanomaterials (Basel, Switzerland), 10(6) (2020-06-19)
Furaneol is a widely used flavoring agent, which can be naturally found in different products, such as strawberries or thermally processed foods. This is why it is extremely important to detect furaneol in the food industry using ultra-sensitive, stable, and
Sensory study on the character impact odorants of roasted Arabica coffee.
Czerny M, et al.
Journal of Agricultural and Food Chemistry, 47(2), 695-699 (1999)
Marije van Beilen et al.
PloS one, 6(9), e23857-e23857 (2011-10-05)
Little is known about the influence of visual characteristics other than colour on flavor perception, and the complex interactions between more than two sensory modalities. This study focused on the effects of recognizability of visual (texture) information on flavor perception
Flavor chemistry of small fruits: blackberry, raspberry, and blueberry.
Du X & Qian M.
ACS Symp. Ser., 1035, 27?43-27?43 (2010)
Isabelle Effenberger et al.
Scientific reports, 9(1), 10943-10943 (2019-07-31)
Glucosyltransferases are versatile biocatalysts to chemically modify small molecules and thus enhance their water solubility and structural stability. Although the genomes of all organisms harbor a multitude of glucosyltransferase genes, their functional characterization is hampered by the lack of high-throughput

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service