263834
Tetrahexylammonium chloride
96%
Synonym(s):
Tetra-n-hexylammonium chloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
[CH3(CH2)5]4N(Cl)
CAS Number:
Molecular Weight:
390.13
Beilstein:
4605091
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
96%
form
powder
mp
111-113 °C (lit.)
SMILES string
[Cl-].CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC
InChI
1S/C24H52N.ClH/c1-5-9-13-17-21-25(22-18-14-10-6-2,23-19-15-11-7-3)24-20-16-12-8-4;/h5-24H2,1-4H3;1H/q+1;/p-1
InChI key
ODTSDWCGLRVBHJ-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- Development of supramolecular solvents: It serves as a key component in the synthesis of ribbon-shaped supramolecular solvents, which are employed for greener microextraction techniques of water-soluble organic pollutants, highlighting its role in environmental chemistry (Algar et al., 2023).
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
A Kaufmann et al.
Analytica chimica acta, 1107, 113-126 (2020-03-24)
Quadrupole based mass spectrometry based detection has experienced enormous improvements in terms of sensitivity over the last centuries. This development has not been equally matched with improvements in selectivity. Hence, the use of unit mass based MS/MS transitions or high
Zeynep S Teksin et al.
Journal of controlled release : official journal of the Controlled Release Society, 116(1), 50-57 (2006-10-20)
Parallel Artificial Membrane Permeability Assay (PAMPA) is a method to screen drug candidates for membrane permeability. The objective was to characterize the transport of a model weak base, metoprolol, across a three lipid-component PAMPA system (denoted A-PAMPA, for anionic-PAMPA) and
The effects of tetrahexylammonium chloride on calcium mobilization from cerebellar microsomes.
L G Sayers et al.
Biochemical Society transactions, 21(2), 105S-105S (1993-05-01)
A M Lisi et al.
Journal of chromatography, 581(1), 57-63 (1992-10-02)
A simple and efficient procedure has been developed for the derivatization of diuretic agents in human urine by direct extractive alkylation and their detection by gas chromatography-mass spectrometry. The procedure is an improvement over previous extractive alkylation methods because of
N Surendran et al.
Journal of chromatography. B, Biomedical sciences and applications, 691(2), 305-312 (1997-04-11)
The objective of this research was to develop a rapid, sensitive and reliable method for the separation of phosphonodipeptide prodrugs and parent compounds to facilitate the evaluation of cell permeation using in vitro cell culture models. Separation was accomplished isocratically
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service