Skip to Content
Merck
All Photos(2)

Key Documents

262269

Sigma-Aldrich

1-Heptene

97%

Synonym(s):

1-n-Heptene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH=CH2
CAS Number:
Molecular Weight:
98.19
Beilstein:
1098332
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
97%

vapor pressure

101 mmHg ( 37.7 °C)

Quality Level

Assay

97%

form

liquid

autoignition temp.

505 °F

refractive index

n20/D 1.400 (lit.)

bp

94 °C (lit.)

mp

−119 °C (lit.)

solubility

water: soluble 0.0182 g/L at 25 °C

density

0.697 g/mL at 25 °C (lit.)

functional group

allyl

SMILES string

CCCCCC=C

InChI

1S/C7H14/c1-3-5-7-6-4-2/h3H,1,4-7H2,2H3

InChI key

ZGEGCLOFRBLKSE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Heptene is an aliphatic hydrocarbon. It is used as a reagent as well as a solvent in organic synthesis. It also acts as an organic additive in some reactions.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

15.8 °F - closed cup

Flash Point(C)

-9.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liquid phase alkylation of toluene with 1-heptene over a HFAU zeolite: evidence for transalkylation between toluene and non-desorbed products.
Da Z, et al.
Applied Catalysis A: General, 182(2), 407-411 (1999)
Ilya Borisov et al.
Polymers, 12(6) (2020-05-30)
For the first time, the effect of the side-chain in polyalkylmethylsiloxane towards pervaporative removal of methyl tert-butyl ether (MTBE) from water was studied. The noticeable enhancement of separation factor during the pervaporation of 1 wt.% MTBE solution in water through
Hiroshi Toda et al.
Applied and environmental microbiology, 81(6), 1919-1925 (2015-01-04)
We describe the development of biocatalysis for producing optically pure straight-chain (S)-epoxyalkanes using styrene monooxygenase of Rhodococcus sp. strain ST-10 (RhSMO). RhSMO was expressed in the organic solvent-tolerant microorganism Kocuria rhizophila DC2201, and the bioconversion reaction was performed in an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service