Skip to Content
Merck
All Photos(1)

Documents

76971

Sigma-Aldrich

1-Pentene

≥98.5% (GC)

Synonym(s):

α-n-Amylene, 1-Methyl-3-butene, 4-Methyl-1-butene, Propylethylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH=CH2
CAS Number:
Molecular Weight:
70.13
Beilstein:
1731629
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

2.4 (vs air)

vapor pressure

10.27 psi ( 20 °C)

Assay

≥98.5% (GC)

form

liquid

autoignition temp.

522 °F

expl. lim.

8.7 %

refractive index

n20/D 1.371 (lit.)
n20/D 1.371

bp

29.9-30.1 °C (lit.)

density

0.641 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCC=C

InChI

1S/C5H10/c1-3-5-4-2/h3H,1,4-5H2,2H3

InChI key

YWAKXRMUMFPDSH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Pentene can be used as a component in gasoline surrogates. It can also be used in the synthesis of (±)-byssochlamic acid and rugulactone.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 1 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-59.8 °F - closed cup

Flash Point(C)

-51 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Total synthesis of (.+-.)-byssochlamic acid.
White, James D et al.
Journal of the American Chemical Society, 114(24), 9673-9674 (1992)
Regioselective Tandem Ring Closing/Cross Metathesis of 1, 5-Hexadien-3-ol Derivatives: Application to the Total Synthesis of Rugulactone.
Cros, Fanny et al.
European Journal of Organic Chemistry, 2010(26), 5063-5070 (2010)
The oxidation of a gasoline surrogate in the negative temperature coefficient region.
Lenhert, David B et al.
Combustion and Flame, 156(3), 549-564 (2009)
Javier A Barrera et al.
Environmental science and pollution research international, 24(33), 26049-26059 (2017-09-25)
Rate coefficients for the gas-phase reactions of OH radicals and Cl atoms with 1-methoxy-2-propanone (1-M-2-PONE), 1-methoxy-2-propanol (1-M-2-POL), and 1-methoxy-2-butanol (1-M-2-BOL) were determined at room temperature and atmospheric pressure using a conventional relative-rate technique. The following absolute rate coefficients were derived:
Yu Shan Susanna Wan et al.
Chemical communications (Cambridge, England), (8)(8), 878-879 (2002-07-19)
A zeolite-based microengineered reactor was fabricated and tested for 1-pentene epoxidation over titanium silicalite-1 (TS-1) catalyst, which has been selectively incorporated within the microreactor channel using a new synthesis procedure.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service