257494
3-Cyano-PROXYL
for ESR-spectroscopy
Synonym(s):
3-Cyano-2,2,5,5-tetramethyl-1-pyrrolidinyloxy, free radical
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C9H15N2O
CAS Number:
Molecular Weight:
167.23
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
grade
for ESR-spectroscopy
Quality Level
form
solid
mp
31-33 °C (lit.)
functional group
nitrile
storage temp.
2-8°C
SMILES string
CC1(C)CC(C#N)C(C)(C)N1[O]
InChI
1S/C9H15N2O/c1-8(2)5-7(6-10)9(3,4)11(8)12/h7H,5H2,1-4H3
InChI key
RQRRZZIMMXPAGX-UHFFFAOYSA-N
General description
Spin label
Application
3-Cyano-PROXYL is a stable free radical that can be used in the ESR technique as a nitroxide standard:
- For the estimation of relative concentrations of the spin adducts of 5,5-dimethyl-1-pyrroline-N-oxide (DMPO) with ·OH and O2.- radicals.
- For the determination of free radicals present in an aqueous cigarette tar extract.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
S M Hahn et al.
Radiation research, 132(1), 87-93 (1992-10-01)
The nitroxide Tempol, a stable free radical, has recently been shown to protect mammalian cells against several forms of oxidative stress including radiation-induced cytotoxicity. To extend this observation, six additional water-soluble nitroxides with different structural features were evaluated for potential
Detection of free radicals in aqueous extracts of cigarette tar by electron spin resonance
Zang L-Y, et al.
Free radical biology & medicine, 19(2), 161-167 (1995)
Scavenging of superoxide anion radical by chaparral
Zang L-Y, et al.
Molecular and Cellular Biochemistry, 196(1-2), 157-161 (1999)
Effect of chlorogenic acid on hydroxyl radical
Zhang L-Y, et al.
Molecular and Cellular Biochemistry, 247(1-2), 205-210 (2003)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service