253324
3-Carboxy-PROXYL
98%
Synonym(s):
3-(Carboxy)-2,2,5,5-tetramethyl-1-pyrrolidinyloxy, 3-Carboxy-2,2,5,5-tetramethyl-1-pyrrolidinyloxy, free radical
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C9H16NO3
CAS Number:
Molecular Weight:
186.23
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
mp
200 °C (dec.) (lit.)
storage temp.
2-8°C
SMILES string
CC1(C)CC(C(O)=O)C(C)(C)N1[O]
InChI
1S/C9H16NO3/c1-8(2)5-6(7(11)12)9(3,4)10(8)13/h6H,5H2,1-4H3,(H,11,12)
InChI key
GEPIUTWNBHBHIO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Spin-label that has been used to study molecular migration and long-term stability of seeds and pollen. Has been covalently attached to oligonucleotides to prepare a DNA probe sensitive to electron spin resonance. The acetoxymethyl ester is an esterase-sensitive probe that crosses the blood-brain barrier for electron spin resonance-computed tomographic imaging.
Spin-label. The anionic form has been used to determine distribution coefficients of neutral solubilizates in anionic micelles.
Used in the preparation of a tetraphenylporphyrin bearing a nitroxyl radical.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
J. Chem. Soc., Faraday, 87, 947-947 (1991)
I Seimenis et al.
Magnetic resonance in medicine, 37(4), 552-558 (1997-04-01)
Proton electron double resonance imaging (PEDRI) was used for monitoring in vivo the distribution, metabolism and, in particular, the excretion mechanism of the exogenous nitroxide free radical proxyl carboxylic acid (PCA) in the rat. PCA clearance half-lives through liver, abdominal
V Quaresima et al.
Physics in medicine and biology, 43(7), 1937-1947 (1998-08-14)
Much outstanding progress concerning the application of ESR spectroscopy/imaging in the biomedical field has been made in recent years. The literature in this field has already been specifically covered by several reviews. The aim of this article is to provide
I Katzhendler et al.
Pharmaceutical research, 17(10), 1299-1308 (2001-01-06)
The present study was conducted in order to investigate the correlation between the hydration properties of HPMC and EA matrices, gel microstructure and mobility, crystalline changes occurring in the gel and CBZ release kinetics. The influence of HPMC and EA
B L Bales et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 132(2), 279-286 (1998-06-20)
A simple expression is derived to compute the total Gaussian linewidth of a Voigt line that is broadened by sinusoidal magnetic-field modulation as follows: delta HPPG(Hm)2 = delta HPPG(0)2 + kappa 2Hm2, where delta HPPG(Hm) is the Gaussian linewidth observed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service