Skip to Content
Merck
All Photos(2)

Key Documents

249955

Sigma-Aldrich

Tin(IV) chloride solution

1.0 M in methylene chloride

Synonym(s):

Stannic chloride, Tin tetrachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SnCl4
CAS Number:
Molecular Weight:
260.52
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

form

liquid

reaction suitability

core: tin
reagent type: catalyst

concentration

1.0 M in methylene chloride

density

1.419 g/mL at 25 °C

SMILES string

Cl[Sn](Cl)(Cl)Cl

InChI

1S/4ClH.Sn/h4*1H;/q;;;;+4/p-4

InChI key

HPGGPRDJHPYFRM-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Tin(IV) chloride shows Lewis acid character and is generally used for the synthesis of SnO2 nanoparticles by sol-gel method.

Application

Tin(IV) chloride solution can be used for chloromethylation reaction of polysulfones to develop polymer electrolyte membranes.

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphoric acid doped polysulfone membranes with aminopyridine pendant groups and imidazole cross-links.
Hink S, et al.
European Polymer Journal, 72(20), 102-113 (2015)
Molecular addition compounds of tin (IV) chloride. I. Interaction with benzonitriles in benzene solution.
Brown TL and Kubota M
Journal of the American Chemical Society, 83(2), 331-334 (1961)
Solid-State and Solution Structural Study of Acetylacetone-Modified Tin (IV) Chloride Used as a Precursor of SnO2 Nanoparticles Prepared by a Sol- Gel Route.
Briois V, et al.
Chemistry of Materials, 16(20), 3885-3894 (2004)
Yasuyuki Kita et al.
The Journal of organic chemistry, 71(14), 5191-5197 (2006-07-01)
The Lewis acid-promoted rearrangement of 2,2,3,3-tetrasubstituted 2,3-epoxy alcohols with several kinds of protecting groups was investigated. When SnCl4 is used as a Lewis acid, the reaction proceeds in a regio- and stereo-controlled manner to afford two types of carbonyl compounds
Gerry A Griffith et al.
Journal of the American Chemical Society, 128(40), 13130-13141 (2006-10-05)
Difluorinated alkenoate ethyl 3,3-difluoro-2-(N,N-diethylcarbamoyloxy)-2-propenoate reacts rapidly and in high yield with furan and a range of substituted furans in the presence of a tin(IV) catalyst. Non-fluorinated congener 2-(N,N-diethylcarbamoyloxy)-2-propenoate fails to react at all under the same conditions. These reactions have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service