Skip to Content
Merck
All Photos(1)

Key Documents

246077

Sigma-Aldrich

Diethyl sebacate

98%

Synonym(s):

Decanedioic acid 1,10-diethyl ester, NSC 8911, Sebacic acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCO(CH2)8COOC2H5
CAS Number:
Molecular Weight:
258.35
Beilstein:
1790779
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.436 (lit.)

bp

312 °C (lit.)

mp

1-2 °C (lit.)

density

0.963 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CCCCCCCCC(=O)OCC

InChI

1S/C14H26O4/c1-3-17-13(15)11-9-7-5-6-8-10-12-14(16)18-4-2/h3-12H2,1-2H3

InChI key

ONKUXPIBXRRIDU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Shoji et al.
The Journal of pharmacy and pharmacology, 46(8), 643-646 (1994-08-01)
The effects of a topical preparation of mycophenolic acid on the experimental allergic contact dermatitis induced by dinitrofluorobenzene was investigated. Visual assessment of skin reactions showed significant efficacy of a topical preparation of mycophenolic acid. This efficacy appeared from the
Allergic contact dermatitis from ethyl sebacate.
Y Kabasawa et al.
Contact dermatitis, 22(4), 226-226 (1990-04-01)
Allergic contact dermatitis due to diethyl sebacate in a hand cream.
Tomohiko Narita et al.
Contact dermatitis, 55(2), 117-117 (2006-08-26)
Allergic contact dermatitis due to diethyl sebacate.
E Sasaki et al.
Contact dermatitis, 36(3), 172-172 (1997-03-01)
B Guieysse et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 52(10-11), 97-105 (2006-02-08)
Two-phase partitioning bioreactors offer many advantages for the removal of toxic pollutants. In particular, such systems can be loaded with very large quantities of pollutants without risks of microbial inhibition, they are self-regulated and they prevent the risks of hazardous

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service