Skip to Content
Merck
All Photos(1)

Key Documents

240249

Sigma-Aldrich

Thiophenol

≥99%

Synonym(s):

Benzenethiol, Phenyl mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SH
CAS Number:
Molecular Weight:
110.18
Beilstein:
506523
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.8 (vs air)

vapor pressure

1.4 mmHg ( 20 °C)

Assay

≥99%

form

liquid

refractive index

n20/D 1.588 (lit.)

bp

169 °C (lit.)

mp

−15 °C (lit.)

solubility

alcohol: very soluble(lit.)
benzene: miscible(lit.)
carbon disulfide: miscible(lit.)
diethyl ether: miscible(lit.)
water: insoluble(lit.)

density

1.073 g/mL at 25 °C (lit.)

SMILES string

Sc1ccccc1

InChI

1S/C6H6S/c7-6-4-2-1-3-5-6/h1-5,7H

InChI key

RMVRSNDYEFQCLF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Thiophenol can be used:
  • To prepare (3H-benzo[c][1,2]diselenol-3-yl)(phenyl)sulfane and β-lactones.
  • As a surface capping ingredient in the preparation Fe-doped ZnS nanoparticles exhibiting ferromagnetic properties.
  • As an oxygen scavenger in the N-deacetylation of chitin to yield chitosan C.
  • As an activator in the coupling reactions of carboxylic acids with isonitriles.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 3 - Repr. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

122.0 °F - closed cup

Flash Point(C)

50 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 56, 1176-1176 (1991)
A practical and efficient method for the synthesis of .beta.-lactones
Danheiser RL and Nowick JS
The Journal of Organic Chemistry, 56(3), 1176-1185 (1991)
Synthesis, structure and reactivity of [o-(2, 6-diisopropylphenyliminomethinyl) phenyl] selenenyl selenocyanate (RSeSeCN) and related derivatives
Rakesh P, et al.
Dalton Transactions, 43(25), 9431-9437 (2014)
Synthesis and characterization of thiophenol passivated Fe-doped ZnS nanoparticles
Sambasivam S, et al.
Materials Science and Engineering, B, 150(2), 125-129 (2008)
Solid state structure of chitosan prepared under different N-deacetylating conditions
Prashanth KVH, et al.
Carbohydrate Polymers, 50(1), 27-33 (2002)

Articles

Markovnikov and anti-Markovnikov alkene reactivity differences are discussed, highlighting challenges and catalytic advancements.

Markovnikov and anti-Markovnikov alkene reactivity differences are discussed, highlighting challenges and catalytic advancements.

Markovnikov and anti-Markovnikov alkene reactivity differences are discussed, highlighting challenges and catalytic advancements.

Markovnikov and anti-Markovnikov alkene reactivity differences are discussed, highlighting challenges and catalytic advancements.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service