Skip to Content
Merck
All Photos(1)

Documents

238414

Sigma-Aldrich

1,3-Diiodopropane

99%, contains copper as stabilizer

Synonym(s):

Trimethylene diiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
I(CH2)3I
CAS Number:
Molecular Weight:
295.89
Beilstein:
1731117
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

contains

copper as stabilizer

refractive index

n20/D 1.642 (lit.)

bp

111-113 °C/31 mmHg (lit.)

density

2.576 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

ICCCI

InChI

1S/C3H6I2/c4-2-1-3-5/h1-3H2

InChI key

AAAXMNYUNVCMCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The adsorption of 1,3-diiodopropane on a molybdenum-aluminum alloy thin film formed on dehydroxylated alumina has been studied.

Application

1,3-Diiodopropane has been used in the preparation of synthetically challenging bridged 1,2,4,5-tetraoxanes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Feng Gao et al.
The journal of physical chemistry. B, 110(25), 12555-12571 (2006-06-28)
The adsorption of C3 hydrocarbons propylene, 1-iodopropane, and 1,3-diiodopropane is studied in ultrahigh vacuum on a molybdenum-aluminum alloy formed by molybdenum hexacarbonyl reaction with a planar alumina film grown on a Mo(100) substrate. Carbon-iodine bond scission occurs below approximately 200
Shyamala Pillai et al.
Journal of fluorescence, 22(4), 1021-1032 (2012-03-28)
A variety of contemporary analytical platforms, utilized in technical and biological applications, take advantage of labeling the objects of interest with fluorescent tracers-compounds that can be easily and sensitively detected. Here we describe the synthesis of new fluorescent quinoline and
Gemma L Ellis et al.
Bioorganic & medicinal chemistry letters, 18(5), 1720-1724 (2008-02-05)
Here we present an efficient route into synthetically challenging bridged 1,2,4,5-tetraoxanes. The key to the success of this route is the use of H(2)O(2) and catalytic I(2) to form the gem-dihydroperoxide followed by a Ag(2)O mediated alkylation using 1,3-diiodopropane. Using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service