125903
1,3-Dibromopropane
ReagentPlus®, 99%
Synonym(s):
Trimethylene dibromide
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
Br(CH2)3Br
CAS Number:
Molecular Weight:
201.89
Beilstein:
635662
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Recommended Products
vapor density
7 (vs air)
Quality Level
product line
ReagentPlus®
Assay
99%
form
liquid
refractive index
n20/D 1.524 (lit.)
bp
167 °C (lit.)
mp
−34 °C (lit.)
density
1.989 g/mL at 25 °C (lit.)
SMILES string
BrCCCBr
InChI
1S/C3H6Br2/c4-2-1-3-5/h1-3H2
InChI key
VEFLKXRACNJHOV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
1,3-Dibromopropane is a dihalogenated propane. It undergoes reduction catalyzed by electrogenerated solution-phase nickel(I) salen and nickel(I) salen confined in a polymer film on the surface of a carbon electrode to afford cyclopropane and propylene, respectively.
Application
1,3-Dibromopropane may be used in the preparation of chiral geminal dicationic ionic liquid and 1,3-bis(1-7′-chloro-4-quinolyl-4- piperazinyl)propane.3
Legal Information
ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 2 - Flam. Liq. 3 - Skin Irrit. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
129.2 °F - closed cup
Flash Point(C)
54 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Synthesis of Chiral Geminal Dicationic Ionic Liquid from Amino Acids.
Yin A, et al.
Asian Journal of Chemistry, 25(12), 6721-6721 (2013)
Catalytic reduction of α,ω-dihaloalkanes with nickel (I) salen as a homogeneous-phase and polymer-bound mediator.
Dahm CE and Peters DG.
Journal of Electroanalytical Chemistry, 406(1), 119-129 (1996)
E E Badr et al.
Journal of oleo science, 59(12), 647-652 (2010-11-26)
A fatty hydrazide based cationic gemini surfactants, 1,3-bis (N'-acyl-N,N-dimethylhydrazinium) propane dibromide which possess hydrolyzable amido moieties in the lipophilic portions, were prepared by reacting 1,3-bromopropane with N,N-dimethyl fatty hydrazide. The surface properties were explained and discussed based on the effect
A Witkowski et al.
The Journal of biological chemistry, 267(26), 18488-18492 (1992-09-15)
Thioesterase II is a 29-kDa monomer which, in certain specialized tissues, acts as a chain terminator in fatty acid synthesis by hydrolyzing medium-chain fatty acids from the fatty acid synthase. As with serine proteases, hydrolysis appears to involve acylation of
S P James et al.
Toxicology letters, 8(1-2), 7-15 (1981-04-01)
Oral administration of 1,3-dibromopropane (2 mmol/kg) to rats resulted in a marked decrease in the level of hepatic glutathione (GSH). Sulphur-containing [14C]-metabolites were excreted in the bile of rats dose with 1,3-bromo[14C]propane and were subjected to enterohepatic cycling. After an
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service