Skip to Content
Merck
All Photos(1)

Key Documents

238368

Sigma-Aldrich

1-Chlorooctadecane

96%

Synonym(s):

Octadecyl chloride, Stearyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)16CH2Cl
CAS Number:
Molecular Weight:
288.94
Beilstein:
1703350
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

refractive index

n20/D 1.451 (lit.)

bp

157-158 °C/1.5 mmHg (lit.)

density

0.849 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCCl

InChI

1S/C18H37Cl/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h2-18H2,1H3

InChI key

VUQPJRPDRDVQMN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Kinetic and thermodynamic analysis of catalytic hydrodechlorination of 1-chlorooctadecane in supercritical carbon dioxide using 5% Pd supported on γ-Al2O3 has been reported. Incorporation of 1-chlorooctadecane into a host monolayer of stearic acid has been reported.

Application

1-Chlorooctadecane has been used as internal standard for the determination of endocrine disrupters in water samples by stir bar sorptive extraction method.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Catalytic hydrodechlorination of 1-chlorooctadecane, 9, 10-dichlorostearic acid, and 12, 14-dichlorodehydroabietic acid in supercritical carbon dioxide.
Aikawa B, et al.
Applied Catalysis. B, Environmental, 43(4), 371-387 (2003)
A Peñalver et al.
Journal of chromatography. A, 1007(1-2), 1-9 (2003-08-20)
Stir bar sorptive extraction (SBSE) combined with gas chromatography (GC) with mass spectrometric detection (MS) has been applied to determine a group of suspected endocrine disrupters in water samples. One centimeter stir bars coated with PDMS were used to extract
The production of stable monolayers from nonamphiphilic 1-chlorooctadecane.
Peters A and Rogers K.
Journal of Colloid and Interface Science, 157(2), 511-512 (1993)
Elisabeth Aubert et al.
Lipids, 39(1), 75-79 (2004-04-02)
The lipids of the gram-negative marine bacterium Marinobacter hydrocarbonoclasticus, cultivated in synthetic seawater supplemented with 1-chlorooctadecane as sole source of carbon, were isolated, purified, and their structures determined. Three pools of lipids were isolated according to the sequential procedure used:
Abid Hussain et al.
Molecules (Basel, Switzerland), 25(8) (2020-04-25)
Date palm dust mites are important pests severely infesting valuable nutritious fruits (dates) of date palm. In search of an alternative to acaricides, joint action of Metarhizium anisopliae EBCL 02049 spores and 1-Chlorooctadecane was evaluated as a potential candidate for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service