
1-Chlorooctadecane

Synonym(s):
Octadecyl chloride, Stearyl chloride, 1-Chlorooctadecane
Linear Formula:
CH3(CH2)17Cl
CAS Number:
Molecular Weight:
288.94
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing