Skip to Content
Merck
All Photos(2)

Key Documents

229725

Sigma-Aldrich

Lead(II) fluoride

99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PbF2
CAS Number:
Molecular Weight:
245.20
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.99% trace metals basis
form:
powder and chunks

Quality Level

Assay

99.99% trace metals basis

form

powder and chunks

reaction suitability

reagent type: catalyst
core: lead

impurities

≤150.0 ppm Trace Metal Analysis

mp

824 °C (lit.)

density

8.445 g/mL at 25 °C (lit.)

SMILES string

F[PbH2]F

InChI

1S/2FH.Pb/h2*1H;/q;;+2/p-2

InChI key

FPHIOHCCQGUGKU-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junjie Zhang et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 62(4-5), 831-834 (2005-08-13)
Up-conversion luminescence properties of a Tm3+/Yb3+ codoped oxyfluoride glass-ceramics under 980 nm excitation are investigated. Intense blue emission centered at 476 nm, corresponding to 1G4-->3H6 transitions of Tm3+ was simultaneously observed in the transparent oxyfluoride glass ceramics at room temperature.
A S Filin et al.
Gigiena i sanitariia, (3)(3), 28-29 (1992-03-01)
Effect of lead fluoride dust on the micronuclear reticulocyte content in the bone marrow was shown. Some functional changes in the testicles and increase in the fetal death rate in exposure to 50 mg/m3 of lead fluoride dust were noted.
Honghan Fei et al.
Journal of the American Chemical Society, 134(26), 10729-10732 (2012-06-20)
We demonstrate the complete exchange of the interlamellar anions of a 2-D cationic inorganic material. The α,ω-alkanedisulfonates were exchanged for α,ω-alkanedicarboxylates, leading to two new cationic materials with the same [Pb(2)F(2)](2+) layered architecture. Both were solved by single crystal X-ray
A I Fadeev
Meditsina truda i promyshlennaia ekologiia, (10)(10), 4-6 (1996-01-01)
The article deals with results obtained in hygienic evaluation of production and application of new monocrystalline compounds based on cesium, sodium, iodine, lead, scandium, fluorine. The evaluation covered work conditions and health state of those engaged into production of monocrystalline
Géraldine Dantelle et al.
Physical chemistry chemical physics : PCCP, 9(41), 5591-5598 (2007-10-25)
beta-PbF(2) single-crystals and nanocrystals in transparent glass-ceramics doped with ErF(3) have been synthesized and studied with two complementary techniques: electron paramagnetic resonance (EPR) and optical spectroscopy (absorption, selective excitation, fluorescence). A comparative study shows that, in both single-crystals and glass-ceramics

Articles

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service