Skip to Content
Merck
All Photos(4)

Documents

223670

Sigma-Aldrich

Bis(cyclopentadienyl)zirconium(IV) chloride hydride

95%

Synonym(s):

Di(cyclopentadienyl)zirconium(IV) chloride hydride, Schwartz’ reagent, Zirconocene chloride hydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11ClZr
CAS Number:
Molecular Weight:
257.87
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder

reaction suitability

core: zirconium
reagent type: catalyst
reagent type: reductant

SMILES string

[H][Zr]Cl.[CH]1[CH][CH][CH][CH]1.[CH]2[CH][CH][CH][CH]2

InChI

1S/2C5H5.ClH.Zr.H/c2*1-2-4-5-3-1;;;/h2*1-5H;1H;;/q;;;+1;/p-1

InChI key

PSYHEBZUPSHSKK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Bis(cyclopentadienyl)zirconium(IV) chloride hydride, also known as Schwartz′s reagent, is utilized as a catalyst in in olefin polymerization.

Application

Bis(cyclopentadienyl)zirconium(IV) chloride hydride can be used:
  • To convert amides to aldehydes.
  • As a reagent for the functionalization of olefins and alkynes.
  • As a reducing agent in the preparation of (E)-vinyl organochalcogenides.
  • To prepare a dioxaborolane derivative, which is a key intermediate for the synthesis of bridged-nicotine analogs.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - Water-react 2

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organic Syntheses, 71, 83-83 (1992)
Preparation, characterization, and activity of silica supported metallocene catalysts
L Britcher, et al.
Chemistry of Materials, 16, 5713-5720 (2004)
Concise synthesis of new bridged-nicotine analogues
Crestey F, et al.
Tetrahedron, 68(5), 1417-1421 (2012)
Bruce Ganem et al.
The Journal of organic chemistry, 72(11), 3981-3987 (2007-03-27)
In this Perspective, which describes the achievements recognized by the 2007 ACS Award for Creative Invention, we discuss the discovery of a new synthetic reaction and its translation into a substantially improved method for manufacturing a major pharmaceutical product--the blockbuster
Paclitaxel from primary taxanes: a perspective on creative invention in organozirconium chemistry
Ganem B and Franke RR
The Journal of Organic Chemistry, 72(11), 3981-3987 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service