Skip to Content
Merck
All Photos(1)

Documents

224308

Sigma-Aldrich

Dilithium tetrachlorocuprate(II) solution

0.1 M in THF

Synonym(s):

Copper dilithium tetrachloride, Dilithium tetrachlorocuprate(2-), Lithium Tetrachlorocuprate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li2CuCl4
CAS Number:
Molecular Weight:
219.24
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

liquid

reaction suitability

reagent type: catalyst
core: copper

concentration

0.1 M in THF

density

0.91 g/mL at 25 °C

SMILES string

[Li+].[Li+].Cl[Cu--](Cl)(Cl)Cl

InChI

1S/4ClH.Cu.2Li/h4*1H;;;/q;;;;+2;2*+1/p-4

InChI key

HCJWWBBBSCXJMS-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst used to couple alkyl halides with alkyl Grignard reagents.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Minamoto et al.
Nucleic acids symposium series, (27)(27), 109-110 (1992-01-01)
The reaction of 1-(2,3-anhydro-5-O-trityl-beta-D-lyxofuranosyl)-2-O-methyluracil (1a) and its thymine analogue (1b) with dilithium tetrahalocuprate (Li2CuX4) revealed excellent to perfect regioselectivity, yielding 2,2'-anhydro-3'-halonucleosides (2a-d), while the same reactions with 2,3-anhydro uracil and thymine nucleosides (4a,b) gave arabinosyl (5a-d) and xylosyl halohydrins (6a-d)
Kenji Mori
Bioscience, biotechnology, and biochemistry, 74(3), 595-600 (2010-03-09)
A mixture of the four stereoisomers of 1,7-dimethylnonyl propanoate, the female sex pheromone of the western corn rootworm (Diabrotica virgifera virgifera LeConte), was synthesized in four different ways by employing one of the following four reactions as the key step:
Tetrahedron, 62, 4851-4851 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service