Skip to Content
Merck
All Photos(4)

Key Documents

221899

Sigma-Aldrich

Vanadium(V) oxide

≥99.6% trace metals basis

Synonym(s):

vanadium, Divanadium pentaoxide, Divanadium pentoxide, Pentaoxodivanadium, Vandia

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
V2O5
CAS Number:
Molecular Weight:
181.88
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.6% trace metals basis

form

powder

reaction suitability

core: vanadium
reagent type: catalyst

mp

690 °C (lit.)

density

3.35 g/mL at 25 °C (lit.)

SMILES string

O=[V](=O)O[V](=O)=O

InChI

1S/5O.2V

InChI key

GNTDGMZSJNCJKK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Vanadium(V) oxide is used in the preparation of vanadium alloys.
  • It is an oxidizing agent, commonly used as a catalyst in contact process (production of sulfuric acid) to oxidize sulfur dioxide to sulfur trioxide.
  • It is widely used as a host material for lithium-ion battery, sodium-ion battery, magnesium-ion battery and pseudocapacitors.
  • V2O5 is also used as the charge injection and extraction materials in organic electronic devices.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 1B - Lact. - Muta. 2 - Repr. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Respiratory system, Respiratory Tract

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vanadium and vanadium compounds.
Ullmann's Encyclopedia of Industrial Chemistry, 1-22 (2000)
Self-assembled vanadium pentoxide (V2O5) hollow microspheres from nanorods and their application in lithium-Ion batteries.
Cao A-M, et al.
Angewandte Chemie (International Edition in English), 44(28), 4391-4395 (2005)
Bilayered vanadium oxide as the host material for reversible beyond lithium ion intercalation.
Clites M, et al.
Advanced Materials Letters, 8(6), 679-688 (2017)
Molecular structure-reactivity relationships for the oxidation of sulfur dioxide over supported metal oxide catalysts.
Dunn JP, et al.
Catalysis Today, 53(4), 543-556 (1999)
T G Folland et al.
Nature communications, 9(1), 4371-4371 (2018-10-24)
Metasurfaces control light propagation at the nanoscale for applications in both free-space and surface-confined geometries. However, dynamically changing the properties of metasurfaces can be a major challenge. Here we demonstrate a reconfigurable hyperbolic metasurface comprised of a heterostructure of isotopically enriched

Articles

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service