Skip to Content
Merck
All Photos(2)

Documents

215821

Sigma-Aldrich

Vanadium(IV) oxide

≥99% trace metals basis

Synonym(s):

Vanadium dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
V2O4
CAS Number:
Molecular Weight:
165.88
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: vanadium

density

4.339 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

O=[V]=O.O=[V]=O

InChI

1S/4O.2V

InChI key

TXPDKHYKECNNNH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Fast-charging high-energy battery-supercapacitor hybrid: anodic reduced graphene oxide-Vanadium (IV) Oxide Sheet-on-Sheet Heterostructure: This study investigates a vanadium(IV) oxide and reduced graphene oxide heterostructure for high-energy, fast-charging battery applications (R Sahoo, et al., 2019).

  • Role of ERK/NFkB in vanadium (IV) oxide mediated osteoblast differentiation in C3H10t1/2 cells: The paper explores the biological effects of vanadium(IV) oxide on osteoblast differentiation, focusing on its potential therapeutic applications (S Srivastava, et al., 2014).

  • Size-dependent catalytic activity of supported vanadium oxide species: oxidative dehydrogenation of propane: This article examines the catalytic properties of vanadium oxide species, particularly their size-dependent activity in the oxidative dehydrogenation of propane (X Rozanska, et al., 2014).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ibrahim Mustafa et al.
Scientific reports, 9(1), 17655-17655 (2019-11-30)
Although free-standing sheets of multiwalled carbon nanotubes (MWCNT) can provide interesting electrochemical and physical properties as electrodes for redox flow batteries, the full potential of this class of materials has not been accessible as of yet. The conventional fabrication methods
Maja Arsic et al.
Plant physiology, 183(4), 1472-1483 (2020-06-17)
Global demand for phosphorus (P) requires new agronomic practices to address sustainability challenges while increasing food production. Foliar P fertilization could increase P use efficiency; however, leaf entry pathways for inorganic phosphate ion (Pi) uptake remain unknown, and it is
Atanu Banerjee et al.
Inorganic chemistry, 59(19), 14042-14057 (2020-09-12)
The synthesis and characterization of one oxidoethoxidovanadium(V) [VVO(L1)(OEt)] (1) and two nonoxidovanadium(IV) complexes, [VIV(L2-3)2] (2 and 3), with aroylhydrazone ligands incorporating naphthalene moieties, are reported. The synthesized oxido and nonoxido vanadium complexes are characterized by various physicochemical techniques, and their
Martin R Otto et al.
Proceedings of the National Academy of Sciences of the United States of America, 116(2), 450-455 (2018-12-28)
We combine ultrafast electron diffraction and time-resolved terahertz spectroscopy measurements to link structure and electronic transport properties during the photoinduced insulator-metal transitions in vanadium dioxide. We determine the structure of the metastable monoclinic metal phase, which exhibits antiferroelectric charge order
Marieke F Jager et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(36), 9558-9563 (2017-08-23)
Coulomb correlations can manifest in exotic properties in solids, but how these properties can be accessed and ultimately manipulated in real time is not well understood. The insulator-to-metal phase transition in vanadium dioxide (VO

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service