219630
DL-2,3-Diaminopropionic acid monohydrochloride
98%
Synonym(s):
3-Amino-DL-alanine monohydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
98%
form
solid
reaction suitability
reaction type: solution phase peptide synthesis
mp
232 °C (dec.) (lit.)
application(s)
peptide synthesis
SMILES string
Cl[H].NCC(N)C(O)=O
InChI
1S/C3H8N2O2.ClH/c4-1-2(5)3(6)7;/h2H,1,4-5H2,(H,6,7);1H
InChI key
SKWCZPYWFRTSDD-UHFFFAOYSA-N
General description
DL-2,3-Diaminopropionic acid monohydrochloride also known as 3-Amino-DL-alanine monohydrochloride, is commonly used in solution phase peptide synthesis.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Amino acids, 43(6), 2537-2543 (2012-06-13)
In this work, we report a synthetic approach to a Fmoc-protected nucleoamino acid, based on L-diaminopropanoic acid, carrying the DNA nucleobase on the alpha-amino group by means of an amide bond, suitable for the solid-phase synthesis of novel nucleopeptides of
Journal of bacteriology, 194(20), 5604-5612 (2012-08-21)
Diaminopropionate ammonia lyase (DAPAL) is a pyridoxal-5'phosphate (PLP)-dependent enzyme that catalyzes the conversion of diaminopropionate (DAP) to pyruvate and ammonia and plays an important role in cell metabolism. We have investigated the role of the ygeX gene of Escherichia coli
Bioorganic & medicinal chemistry letters, 22(17), 5635-5638 (2012-08-04)
Here we report a novel class of peptides-d-diaminopropionic acids (Dap)-for gene delivery. These peptides have attractive properties for gene delivery, and the advantage that they can be easily manipulated in relation to their composition, abiding with tailored-design. We characterized the
Molecular microbiology, 74(3), 594-608 (2009-09-25)
Siderophores are iron-scavenging molecules produced by many microbes. In general, they are synthesized using either non-ribosomal peptide synthetase (NRPS) or NRPS-independent siderophore (NIS) pathways. Staphylococcus aureus produces siderophores, of which the structures of staphyloferrin A and staphyloferrin B are known.
FEBS letters, 582(20), 3125-3131 (2008-08-12)
Zwittermicin A (ZwA) is a hybrid polyketide-non-ribosomal peptide that is thought to be biosynthesized from five proposed building blocks, including the 2,3-diaminopropionate. Candidate genes for de novo biosynthesis of 2,3-diaminopropionate, zwa5A and zwa5B, have been identified in a previous study.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service