Skip to Content
Merck
All Photos(6)

Documents

O2375

Sigma-Aldrich

L-Ornithine monohydrochloride

≥99%

Synonym(s):

(S)-2,5-Diaminopentanoic acid monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12N2O2 · HCl
CAS Number:
Molecular Weight:
168.62
Beilstein:
3625847
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

L-Ornithine monohydrochloride, ≥99%

Quality Level

Assay

≥99%

form

powder

impurities

ammonia and citrulline, essentially free

color

white

application(s)

cell analysis

SMILES string

Cl.NCCC[C@H](N)C(O)=O

InChI

1S/C5H12N2O2.ClH/c6-3-1-2-4(7)5(8)9;/h4H,1-3,6-7H2,(H,8,9);1H/t4-;/m0./s1

InChI key

GGTYBZJRPHEQDG-WCCKRBBISA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Ornithine monohydrochloride has been used to study the blocking effect of L-ornithine on the transport system that carries L-arginine for the overproduction of nitric oxide during lipopolysaccharide induced endotoxemia in rat model. It has also been used in decarboxylation test of amino acids.

Biochem/physiol Actions

L-Ornithine is a product of arginase degradation and plays an important role in ammonia metabolism via the urea cycle. In mammals, ornithine serves as a precursor for polyamines and proline biosynthesis. Spermine synthesized from ornithine, is an abundant polyamine observed in tumors such as breast carcinomas. Administration of L-ornithine is known to improve subjective index of sleep quality. L-ornithine reduces the cortisol/DHEA-S (dehydroepiandrosterone) ratio, that is associated with stress. In mice, L-ornithine is found to affect the levels of plasma growth hormone, corticosterone and melatonin levels in mice.

Other Notes

Product of arginine degradation by arginase

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L-Ornithine affects peripheral clock gene expression in mice
Takafumi Fukuda
Scientific Reports (2016)
THE EFFECT OF L-ORNITHINE ON THE INNER EAR OF RATS IN THE LIPOPOLYSACCHARIDE INDUCED ENDOTOXEMIA MODEL
Keal Gorur
Kulak Burun bo?az Ihtisas Dergisi : KBB = Journal of Ear, Nose, and Throat (2018)
Arginase Activity in Human Breast Cancer Cell Lines: N?-Hydroxy-l-arginine Selectively Inhibits Cell Proliferation and Induces Apoptosis in MDA-MB-468 Cells
Rajan Singh
Cancer Research, 60(12) (2000)
Identification of bacteria potentially producing biogenic amines isolated from Venezuelan white cheese
Alba Lorena BENAVIDES Sierra
Revista Brasileira de Farmacognosia (2016)
Sherry Dadsetan et al.
Biochemical pharmacology, 85(1), 115-123 (2012-10-30)
Combined administration of ornithine and phenylacetate (OP) is proposed as a novel treatment of hyperammonemia and hepatic encephalopathy. Ornithine is believed to increase ammonia fixation into glutamine in muscle tissue and glutamine is subsequently thought to react with phenylacetate forming

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service