Skip to Content
Merck
All Photos(4)

Key Documents

219584

Sigma-Aldrich

2,6-Di-tert-butylpyridine

≥97%

Synonym(s):

2,6-Bis(1,1-dimethylethyl)pyridine, 2,6-Ditert-butylpyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H21N
CAS Number:
Molecular Weight:
191.31
Beilstein:
125886
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97%

form

liquid

refractive index

n20/D 1.473 (lit.)

bp

100-101 °C/23 mmHg (lit.)

density

0.852 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(C)c1cccc(n1)C(C)(C)C

InChI

1S/C13H21N/c1-12(2,3)10-8-7-9-11(14-10)13(4,5)6/h7-9H,1-6H3

InChI key

UWKQJZCTQGMHKD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Reactivity of 2,6-di-tert-butylpyridine with iron(III) tetraphenylporphyrin pi-cation radical has been examined by proton NMR spectroscopy. Reaction of 2,6-di-tert-butylpyridine with methyl iodide and methyl fluorosulfonate under high pressure has been reported.

Application

2,6-Di-tert-butylpyridine was used as proton trapping agent to investigate the living polymerization of isobutylene. It was also used with cerric ammonium nitrate in the α-enolation of aldehydes leading to 1,4-dicarbonyl systems.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective organocatalytic singly occupied molecular orbital activation: the enantioselective alpha-enolation of aldehydes.
Hye-Young Jang et al.
Journal of the American Chemical Society, 129(22), 7004-7005 (2007-05-15)
Karolina A Tarach et al.
ChemSusChem, 12(3), 633-638 (2018-12-07)
A new theoretical and practical framework has been developed through operando study of the zeolite catalytic cracking of low-density polyethylene (as a model reaction) under reaction conditions. Results show that microporous ZSM-5 gives rise to less cracking products. Hierarchical ZSM-5
Reactions of crowded molecules under high pressure. Reactions of 2, 6-di-tert-butylpyridine with methyl iodide and methyl fluorosulfonate under high pressure.
Okamoto Y and Lee KI.
Journal of the American Chemical Society, 97(!4), 4015-4018 (1975)
J. Macromol. Sci., Pure Appl. Chem., A29, 639-639 (1992)
Krystyna Rachlewicz et al.
Inorganic chemistry, 35(5), 1136-1147 (1996-02-28)
The reactivity of iron(III) tetraphenylporphyrin pi-cation radical (TPP(*))Fe(III)(ClO(4))(2), (1-1) iron(III) tetra-p-tolylporphyrin pi-cation radical (TTP(*))Fe(III)(ClO(4))(2) (1-2) and iron(III) tetramesitylporphyrin pi-cation radical (TMP(*))Fe(III)(ClO(4))(2) (1-3) complexes with 2,4,6-collidine, 2,3,6-collidine, 2-picoline, 2,6-di-tert-butylpyridine, and 2,6-dibromopyridine has been examined by (1)H NMR spectroscopy in dichloromethane-d(2) solution

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service