Skip to Content
Merck
All Photos(2)

Key Documents

20430

Sigma-Aldrich

Boc-OSu

≥98.0% (CHN), for peptide synthesis

Synonym(s):

N-(tert-Butoxycarbonyloxy)succinimide, tert-Butyl N-succinimidyl carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13NO5
CAS Number:
Molecular Weight:
215.20
Beilstein:
1536756
EC Number:
MDL number:
UNSPSC Code:
12352005
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

product name

Boc-OSu, ≥98.0% (CHN)

Quality Level

Assay

≥98.0% (CHN)

form

solid

mp

~95 °C (dec.)

application(s)

peptide synthesis

functional group

imide

storage temp.

−20°C

SMILES string

CC(C)(C)OC(=O)ON1C(=O)CCC1=O

InChI

1S/C9H13NO5/c1-9(2,3)14-8(13)15-10-6(11)4-5-7(10)12/h4-5H2,1-3H3

InChI key

VTGFSVGZCYYHLO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Boc-protecting group reagent

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. Franke et al.
Tetrahedron Letters, 4765-4765 (1966)
Stabilization of Bacillus subtilis -amylase by amino group acylation.
J Hora
Biochimica et biophysica acta, 310(1), 264-267 (1973-05-17)
H. Gross et al.
Justus Liebigs Annalen der Chemie, 725, 212-212 (1969)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service