Skip to Content
Merck
All Photos(2)

Documents

204846

Sigma-Aldrich

Ammonium metavanadate

99.95% trace metals basis

Synonym(s):

Ammonium vanadate(V)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4VO3
CAS Number:
Molecular Weight:
116.98
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.95% trace metals basis

form

crystalline

impurities

≤150.0 ppm Trace Metal Analysis

density

2.32 g/cm3 at 25 °C (lit.)

SMILES string

N.O[V](=O)=O

InChI

1S/H3N.H2O.2O.V/h1H3;1H2;;;/q;;;;+1/p-1

InChI key

YBVKNHXQSRDWAA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Ammonium metavanadate is widely used to fabricate cathode material for Li-ion batteries.

It can also be used as a precursor to synthesize various vanadium catalysts such as vanadium carbide and nitride catalysts for hydrogenation, hydrodesulfurization, and hydrodenitrogenation reactions.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 2 - STOT RE 1 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yukti Arora et al.
Scientific reports, 8(1), 12752-12752 (2018-08-26)
Storage of solar radiation is currently accomplished by coupling two separate devices, one that captures and converts the energy into an electrical impulse (a photovoltaic cell) and another that stores this electrical output (a battery or a supercapacitor electrochemical cell).
Ahlem Soussi et al.
Comptes rendus biologies, 329(10), 775-784 (2006-10-10)
Transitional metals, as vanadium, are known to exert noxious effects by generating oxidative stress. Addition of antioxidants in the diet could decrease the cytotoxic effect related to the oxidative stress. The present study, carried out in Wistar rats, is a
Ashraf M Morgan et al.
Pharmacological research, 47(1), 75-85 (2003-01-16)
Vanadium is a ubiquitous trace metal present in most plant and animal tissues. Environmental exposure to trivalent and pentavalent inorganic vanadium compounds has been related to impaired different phases of reproduction. Therefore, the effects of a pentavalent inorganic vanadium compound
Mansour S Abdul Galil et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 67(1), 76-82 (2006-08-22)
A rapid, simple, and most economical spectrophotometric method was proposed for the determination of nitrite in various water samples, soil samples, and roots of leguminous plants. The method is based on decolorizing effect of nitrite on complex formed between hydrogen
I V Victorov et al.
Brain research. Brain research protocols, 5(2), 135-139 (2000-04-25)
Acidophilia is one of the hallmarks of acute neuronal damage and death in brain ischemia, excitotoxic and traumatic lesions and epileptic seizures. We here describe a novel and simple method for visualizing acidophilic neurons on paraffin sections, using vanadium acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service