Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ammonium metavanadate
Synonym(s):
Ammonium vanadate(V)
Linear Formula:
NH
4
VO
3
CAS Number:
7803-55-6
Molecular Weight:
116.98
EC Number:
232-261-3
Products
Product Category
salts (3)
Brand
Sigma-Aldrich (4)
Formula Weight
Physical Form
Purity
Quality Segment
Reagent Type
Core
Special Grade
ACS reagent (2)
Puriss (1)
Puriss P.A. (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
398128
Ammonium metavanadate,
ACS reagent, ≥99.0%
Expand
View Pricing
10028
Ammonium metavanadate,
puriss. p.a., ACS reagent, ≥99.0% (RT)
Expand
View Pricing
205559
Ammonium metavanadate,
99%
Expand
View Pricing
204846
Ammonium metavanadate,
99.95% trace metals basis
Expand
View Pricing