Skip to Content
Merck
All Photos(1)

Documents

201618

Sigma-Aldrich

Sudan Red 7B

Synonym(s):

Fat Red 7B, Fat red bluish, Solvent Red 19

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H21N5
CAS Number:
Molecular Weight:
379.46
Colour Index Number:
26050
Beilstein:
3534323
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

mp

130 °C (dec.) (lit.)

solubility

acetone: 10 mg/mL

λmax

364 nm (2nd)
533 nm

ε (extinction coefficient)

≥14230 at 367-377 nm in toluene
≥24660 at 528-538 nm in toluene

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CCNc1ccc2ccccc2c1\N=N\c3ccc(cc3)\N=N\c4ccccc4

InChI

1S/C24H21N5/c1-2-25-23-17-12-18-8-6-7-11-22(18)24(23)29-28-21-15-13-20(14-16-21)27-26-19-9-4-3-5-10-19/h3-17,25H,2H2,1H3/b27-26+,29-28+

InChI key

VKWNTWQXVLKCSG-ZDXBJJIESA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sudan Red 7B has been used to stain the fat bodies of Ostrinia nubilalis (Hübner) larvae.

Biochem/physiol Actions

Sudan red 7B, also known as fat red, is a non-fluorescent stain. It is effectively used to stain lipids and is used to indicate the presence of suberin in cell walls. Sudan red 7B acts as a marker for formosan subterranean termite, Coptotermes fornosanus Shiraki.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A.L. Smit
Root Methods: A Handbook (2013)
NAN-YAO SU
Material und Organismen: Materials and organisms. Materiaux et organismes (1980)
James L. Brewster
Onions and Other Vegetable Alliums (2008)
Effect of Age and Mating Status on Adult European Corn
Borer (Lepidoptera: Crambidae) Dispersal from Small-Grain
Aggregation Plots
Reardon B J
Journal of Economic Entomology, 100, 1116-1123 (2007)
W.E.G. Muller and H.C. Schroder
Biological Response Modifiers ? Interferons, Double-Stranded RNA and 2?,5?-Oligoadenylates (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service