Skip to Content
Merck
All Photos(1)

Key Documents

567517-U

Supelco

Discovery® HS F5 (5 µm) HPLC Columns

L × I.D. 25 cm × 4.6 mm, HPLC Column

Synonym(s):

Discovery Pentafluorophenyl PFP HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Product Name

Discovery® HS F5 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm

material

stainless steel column

Quality Level

Agency

suitable for USP L43

product line

Discovery®

feature

endcapped

manufacturer/tradename

Discovery®

packaging

1 ea of

extent of labeling

12% Carbon loading

parameter

≤70 °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

25 cm × 4.6 mm

surface area

300 m2/g

surface coverage

4 μmol/m2

impurities

<10 ppm metals

matrix

silica gel, high purity, spherical particle platform
fully porous particle

matrix active group

PFP (pentafluorophenyl) phase

particle size

5 μm

pore size

120 Å

operating pH range

2-8

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Guidelines for transferring a C18 method to Discovery® HS F5:
Generally, bases are retained longer on the HS F5 than on a C18. Increasing the organic content of a C18 separation 5 to 10 percent will generally provide similar retention on an HS F5. Results with other compounds are highly variable. However, it is generally true that solutes with log Po/w values less than 2.5 will be retained longer on HS F5 compared to a C18. The degree of difference is highly solute dependent.
The Discovery® HS F5 bonded phase provides reversed-phase separations that are distinctly different from C18 columns. However, compounds will generally elute within the same retention time window, making most C18 methods easily transferable.

Application


  • Determination of 6-hydroxy-1-methyl-1,2,3,4-tetrahydro-beta-carboline, 5-hydroxytryptamine and 5-hydroxyindole acetic acid in the neonatal rat brains using high performance liquid chromatography-electrochemical detection.: This study highlights the use of the Discovery® HS F5 HPLC Column for the sensitive detection of neuroactive compounds in neonatal rat brain samples, advancing our understanding of neurological development and neurotransmitter dynamics (Mao et al., 2009).

  • Advantages of pentafluorophenylpropyl stationary phase over conventional C18 stationary phase--application to analysis of triamcinolone acetonide.: This research showcases the superior performance of the Discovery® HS F5 HPLC Column in analyzing corticosteroid formulations, providing insights that could lead to improved analytical methodologies in pharmaceutical analysis (Havlikova et al., 2008).

  • Determination of ephedrine alkaloids in Ephedra natural products using HPLC on a pentafluorophenylpropyl stationary phase.: Utilizes the Discovery® HS F5 HPLC Column for the precise and efficient separation of ephedrine alkaloids, contributing to safer and more effective use of Ephedra in herbal supplements (Pellati and Benvenuti, 2008).

  • Fast high-performance liquid chromatography analysis of phenethylamine alkaloids in Citrus natural products on a pentafluorophenylpropyl stationary phase.: Demonstrates the efficiency of the Discovery® HS F5 HPLC Column in the rapid analysis of phenethylamine alkaloids, enhancing the quality control of Citrus-based dietary supplements (Pellati and Benvenuti, 2007).

  • High-performance liquid chromatographic method with UV photodiode-array, fluorescence and mass spectrometric detection for simultaneous determination of galantamine and its phase I metabolites in biological samples.: This study uses the Discovery® HS F5 HPLC Column to simultaneously quantify galantamine and its metabolites, aiding in the pharmacokinetic profiling of this Alzheimer′s disease medication (Malakova et al., 2007).

Features and Benefits

  • Unique selectivity
  • Similar retention to C18 (sometimes requires stronger mobile phase)
  • Excellent peak shape
  • Stable, low-bleed LC-MS separations
  • Scalable separations from 3 to 10μm particle sizes

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Umberto M Battisti et al.
Journal of chromatography. A, 1443, 152-161 (2016-03-30)
A "heart-cut" two-dimensional achiral-chiral liquid chromatography triple-quadrupole mass spectrometry method (LC-LC-MS/MS) was developed and coupled to in vivo cerebral microdialysis to evaluate the brain response to the chiral compound (±)-7-chloro-5-(3-furanyl)-3-methyl-3,4-dihydro-2H-1,2,4-benzothiadiazine-1,1-dioxide ((±)-1), a potent positive allosteric modulator (PAM) of AMPA receptor.
Jennie Wang et al.
Journal of pharmaceutical and biomedical analysis, 124, 267-273 (2016-03-13)
An HPLC method for the assay of a DNA topoisomerase inhibitor, LMP776 (NSC 725776), has been developed and validated. The stress testing of LMP776 was carried out in accordance with International Conference on Harmonization (ICH) guidelines Q1A (R2) under acidic
Jana Maláková et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 853(1-2), 265-274 (2007-04-10)
Galantamine, an alkaloid isolated from the bulbs and flowers of Caucasian snowdrop (Galanthus woronowii, Amaryllidaceae) and related species, is employed in human medicine for the treatment of various neuromuscular and neurodegenerative diseases. After the administration, the products of oxidative biotransformation
Lucie Havlíková et al.
Talanta, 76(3), 597-601 (2008-07-01)
A pentafluorophenylpropyl (PFPP) stationary phase was for the first time tested for the simultaneous determination of triamcinolone acetonide, its degradation product triamcinolone and two preservatives, methylparaben, and propylparaben. A new simple isocratic reversed phase HPLC method with UV detection, using
Koji Baba et al.
Journal of chromatography. A, 1354, 109-116 (2014-06-21)
Arsenic species, including arsenous acid, arsenic acid, methylarsonic acid, and dimethylarsinic acid, were determined using HPLC-ICPMS. The species were separated with a Discovery HS F5 column and a simple, volatile, and isocratic mobile phase of 0.1% (v/v) formic acid and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service