Skip to Content
Merck
All Photos(1)

Key Documents

175765

Sigma-Aldrich

tert-Butanol-OD

99 atom % D

Synonym(s):

2-Methyl-2-propan(ol-d), tert-Butan(ol-d), tert-Butyl alcohol-d

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COD
CAS Number:
Molecular Weight:
75.13
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % D

Quality Level

Assay

99% (CP)

form

liquid

refractive index

n20/D 1.3847 (lit.)

bp

83 °C (lit.)

mp

23-26 °C (lit.)

density

0.786 g/mL at 25 °C

mass shift

M+1

SMILES string

[2H]OC(C)(C)C

InChI

1S/C4H10O/c1-4(2,3)5/h5H,1-3H3/i5D

InChI key

DKGAVHZHDRPRBM-UICOGKGYSA-N

Application

tert-Butanol-OD is a stable isotope that can be used:
  • As a model compound in the study of alcohol reactivities on MoVI and WVI oxide clusters.
  • In the synthesis of 8-deuteriocamphor.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of camphor-1, 4-homoenol (7-hydroxy-1, 2-dimethyl [3, 3, 0, 02, 7] tricyclooctane)
Dadson WM and Money T
Canadian Journal of Chemistry, 58(23), 2524-2526 (1980)
Oxidation, reduction, and condensation of alcohols over (MO3) 3 (M= Mo, W) nanoclusters
Fang Z, et al.
The Journal of Physical Chemistry C, 118(39), 22620-22634 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service