Skip to Content
Merck
All Photos(2)

Key Documents

8.22264

Supelco

tert-Butanol

EMPLURA®

Synonym(s):

tert-Butanol, tert-Butyl alcohol, 2-Methyl-2-propanol, Trimethylcarbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COH
CAS Number:
Molecular Weight:
74.12
MDL number:
UNSPSC Code:
12352104
EC Index Number:
200-889-7
NACRES:
NB.21
bp:
81-83 °C/1013 hPa
vapor pressure:
40.7 hPa ( 20 °C)
Pricing and availability is not currently available.

vapor pressure

40.7 hPa ( 20 °C)

Quality Level

product line

EMPLURA®

Assay

≥99% (GC)

form

solid

autoignition temp.

490 °C

expl. lim.

2.3-8.0 % (v/v)

dilution

(low-cost alternative, for basic applications)

impurities

≤0.1% Water

pH

7 (20 °C)

bp

81-83 °C/1013 hPa

mp

24-25 °C

transition temp

flash point 14 °C

density

0.78 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OC(C)(C)C

InChI

1S/C4H10O/c1-4(2,3)5/h5H,1-3H3

InChI key

DKGAVHZHDRPRBM-UHFFFAOYSA-N

General description

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.

Application


  • Elucidating and Tuning Catalytic Sites on Zirconium- and Aluminum-Containing Nodes: Investigates the application of tert-butanol in the tuning of catalytic sites within metal-organic frameworks, essential for advancements in material science and catalysis (Yang & Gates, 2021).

  • Photocatalytic mechanism and performance of a novel wool flake-BiFeO3nanosheet-TiO2 core-shell-structured composite photocatalyst: Describes how tert-butanol acts as a solvent in the synthesis of complex photocatalysts, enhancing their efficiency in environmental cleanup applications (Chen et al., 2021).

  • Effect of cross-linked enzyme aggregate strategy on characterization of sn-1,3 extracellular lipase: Demonstrates the role of tert-butanol in enzyme immobilization techniques to improve the stability and activity of lipases, vital for biotechnological applications (Zhu et al., 2021).

Analysis Note

Purity (GC): ≥ 99.0 %
Identity (IR): conforms
Melting point: 24.0 - 26.0 °C
Water: ≤ 0.10 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

59.0 °F - closed cup

Flash Point(C)

15 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service