Skip to Content
Merck
All Photos(1)

Key Documents

174866

Sigma-Aldrich

1,12-Dibromododecane

98%

Synonym(s):

Dodecamethylene dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH2)12Br
CAS Number:
Molecular Weight:
328.13
Beilstein:
1742763
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

bp

215 °C/15 mmHg (lit.)

mp

38-42 °C (lit.)

SMILES string

BrCCCCCCCCCCCCBr

InChI

1S/C12H24Br2/c13-11-9-7-5-3-1-2-4-6-8-10-12-14/h1-12H2

InChI key

ZJJATABWMGVVRZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,12-Dibromododecane was used in the preparation of tboc-protected ionenes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sean M Ramirez et al.
Macromolecular bioscience, 9(11), 1127-1134 (2009-07-29)
Novel tboc-protected ionenes with M(w) exceeding 30 kDa were prepared from the step-growth polymerization of tert-butyl bis[3-(dimethylamino)propyl]carbamate and 1,12-dibromododecane. The protected ionenes yielded pH-sensitive, protonatable ionenes with pK(a) approximately 6.6 for the conjugate acid of the protonated secondary amine. Polyplexes
Mojca Seručnik et al.
The journal of physical chemistry. B, 122(21), 5381-5388 (2018-01-26)
Complexes of polycations and DNA, also known as polyplexes, have been extensively studied in the past decade, as potential gene delivery systems. Their stability depends strongly on the characteristics of the polycations, as well as the nature of the added
Daria Szymaniak et al.
ChemPlusChem, 85(10), 2281-2289 (2020-09-23)
The synthesis and characteristics are presented of novel double-salt herbicidal ionic liquids (DSHILs) that contain 4-chloro-2-methylphenoxyacetate and trans-cinnamate anions. In the designed synthesis, an anion of natural origin and a herbicidal anion were combined with an amphiphilic bisammonium cation to
María Cordero-Vaca et al.
Analytical and bioanalytical chemistry, 407(16), 4615-4627 (2015-05-01)
Four different crosslinked polymeric ionic liquid (PIL)-based sorbent coatings were evaluated in an automated direct-immersion solid-phase microextraction method (automated DI-SPME) in combination with gas chromatography (GC). The crosslinked PIL coatings were based on vinyl-alkylimidazolium- (ViCnIm-) or vinylbenzyl-alkylimidazolium- (ViBzCnIm-) IL monomers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service