Skip to Content
Merck
All Photos(1)

Key Documents

361283

Sigma-Aldrich

1,8-Dichlorooctane

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl(CH2)8Cl
CAS Number:
Molecular Weight:
183.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.459 (lit.)

bp

115-116 °C/11 mmHg (lit.)

density

1.025 g/mL at 25 °C (lit.)

SMILES string

ClCCCCCCCCCl

InChI

1S/C8H16Cl2/c9-7-5-3-1-2-4-6-8-10/h1-8H2

InChI key

WXYMNDFVLNUAIA-UHFFFAOYSA-N

General description

1,8-Dichlorooctane is a chloroparaffin. The non-exponentiality of proton spin-lattice relaxation in polycrystalline 1,8-dichlorooctane in Zeeman-quadrupole level-crossing experiments at various frequencies is reported. It has important synthetic applications in food, pharmaceutical, biomedical, electrical and electronic sectors.

Application

1,8-Dichlorooctane may be used:
  • as adsorbate to investigate the performance of carbonaceous materials for the adsorption of emerging pollutants
  • to prepare the C20-backbone of 1,20-substituted eicosanes carrying phosphate headgroups and readily derivatizable thiol, maleimido, and activated carboxylic ester moieties
  • in the synthesis of planar-chiral imidazolium salt
  • in the preparation of biosensors
  • in the fabrication of strong polymeric membranes for concentrating amino acids, having applications in food and pharmaceutical industry
  • in the synthesis of perfluorocarbons having biomedical applications

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

244.4 °F

Flash Point(C)

118 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vibrational analysis of 1, 8-dichlorooctane.
Singh D, et al.
Indian Journal of Pure and Applied Physics, 47(10), 696-696 (2009)
Limited spin diffusion towards quadrupolar relaxation centers in 1, 8-dichlorooctane
Kozak A, et al.
Chemical Physics Letters, 74(1), 173-175 (1980)
Yolanda Patiño et al.
Chemosphere, 119 Suppl, S124-S130 (2014-06-09)
The adsorption of three representative emerging pollutants over different kinds of carbonaceous adsorbents has been studied in this work. The adsorbates were nalidixic acid (NAL, representative of a pharmaceutical), 1,8-dichlorooctane (DCO, a chloroparaffin) and methyl-phenoxy-ethanol (MPET, a surfactant). Activated carbons
Synthesis and properties of a diastereopure ionic liquid with planar chirality.
Ishida Y, et al.
Tetrahedron Letters, 47(45), 7973-7976 (2006)
Istvan Jablonkai et al.
Chemistry and physics of lipids, 133(1), 103-112 (2004-12-14)
New 1,20-substituted eicosanes carrying phosphate headgroups and readily derivatizable thiol, maleimido, and activated carboxylic ester moieties were prepared. The C20-backbone of these molecules was assembled by a halopolycarbon homologation from 1,8-dichlorooctane and 1,6-dibromohexane. 20-Mercapto- and 20-maleimido-icosylphosphates were synthesized via omega-bromo

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service