Skip to Content
Merck
All Photos(2)

Key Documents

157872

Sigma-Aldrich

1,3-Dithiane

97%

Synonym(s):

m-Dithiane (7CI), m-Dithiane (8CI)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8S2
CAS Number:
Molecular Weight:
120.24
Beilstein:
102534
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

52-54 °C (lit.)

SMILES string

C1CSCSC1

InChI

1S/C4H8S2/c1-2-5-4-6-3-1/h1-4H2

InChI key

WQADWIOXOXRPLN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,3-Dithiane, a protected formaldehyde anion equivalent, serves as useful labeled synthon.

Application

1,3-Dithiane was used as reagent for deoxygenation of sulfoxides to their corresponding sulfides.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Al-Monsur Jiaul Haque et al.
Chemical communications (Cambridge, England), (32)(32), 4865-4867 (2009-08-05)
We report the use of 1,3-dithiane combined with aryldiazonium cation for the immobilization of biomolecules based on electrochemical addressing.
Yuncong Chen et al.
Chemical communications (Cambridge, England), 48(42), 5094-5096 (2012-04-20)
A novel sensitive and specific Hg(2+) chemodosimeter, derived from 1',3'-dithiane-substituted 2,1,3-benzoxadiazole, displays "turn-on" fluorescent and colorimetric responses via an Hg(2+)-triggered aldehyde recovery reaction. Its potential to monitor Hg(2+) in living organisms has been demonstrated using zebrafish larvae.
A I Noskov et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 77(1), 6-10 (2010-07-17)
The IR spectra of 1,3-dithiane-1-oxide (I) and 1,3-dithia-1-oxocyclohept-5-ene (II) were recorded in solution, solid and liquid phase over 4000-400 cm(-1) spectral range. It was found that both (I) and (II) in liquid phase and solutions exist in two conformations: (I)
Rodolfo A Martinez et al.
Journal of labelled compounds & radiopharmaceuticals, 57(5), 338-341 (2014-05-28)
The 1,3-dithiane is a protected formaldehyde anion equivalent that could serve as a useful labeled synthon. We report a facile synthesis of 1,3-[2-(13)C]- and 1,3-[2-(13)C, 2-(2)H2]dithiane in two steps from [(13)C]- or [(13) C, (2)H3 ]methyl phenyl sulfoxide. We have
Valerie J Peterson et al.
The Biochemical journal, 362(Pt 1), 173-181 (2002-02-07)
Apo and holo forms of retinoic acid receptors, and other nuclear receptors, display differential sensitivity to proteolytic digestion that likely reflects the distinct conformational states of the free and liganded forms of the receptor. We have developed a method for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service