Skip to Content
Merck
All Photos(1)

Key Documents

131970

Sigma-Aldrich

1,4-Thioxane

98%

Synonym(s):

1,4-Oxathiane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8OS
CAS Number:
Molecular Weight:
104.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.5095 (lit.)

bp

147 °C/755 mmHg (lit.)

density

1.114 g/mL at 25 °C (lit.)

SMILES string

C1CSCCO1

InChI

1S/C4H8OS/c1-3-6-4-2-5-1/h1-4H2

InChI key

JBYHSSAVUBIJMK-UHFFFAOYSA-N

Application

1,4-Thioxane (1,4-oxathiane) was used to study the growth and characterization of the vertically aligned zinc oxide (ZnO) nanowires.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

107.6 °F - closed cup

Flash Point(C)

42 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michał Czub et al.
Marine environmental research, 161, 105077-105077 (2020-08-28)
Sulphur mustard (HD) was the most widely produced chemical warfare agent (CWA) in the history of chemical warfare (CW). Simultaneously, the loads of HD account as by far the largest fraction of the sea-dumped CW. Nowadays its presence in the
M W Ellzy et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 57(12), 2417-2432 (2002-01-05)
We present a detailed analysis of the structure and infrared spectra of 1,4-thioxane. The vibrational frequencies of the 1,4-thioxane molecule were analyzed using standard quantum chemical techniques. Frequencies were calculated at the MP2 and DFT levels of theory using the
Xin Zhang et al.
Environmental science & technology, 43(3), 610-615 (2009-02-28)
The long-term fate of chemical weapon debris disposed of in the ocean some 50 years ago, now sinking into marine sediments and leaking into the ocean environment, is poorly known. Direct evidence exists showing chemical weapon agents actively being released
S Hindley et al.
Journal of nanoscience and nanotechnology, 11(9), 8294-8301 (2011-11-22)
Vertically aligned zinc oxide (ZnO) nanowires (NWs) have been grown by liquid injection Metal Organic Chemical Vapour Deposition, using oxygen donor adducts of Me2Zn. The growth and characterisation of the nanowires grown using [Me2Zn(L)] where L = monodentate ethers, tetrahydrofuran
Heidi Lees et al.
Electrophoresis, 38(7), 1075-1082 (2017-01-04)
In this study, novel, fast, and simple methods based on RP-HPLC and MEKC with DAD are developed and validated for the qualitative and quantitative determination of five cyclic sulfur mustard (HD) degradation products (1,4-thioxane, 1,3-dithiolane, 1,4-dithiane, 1,2,5-trithiepane, and 1,4,5-oxadithiepane) in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service