Skip to Content
Merck
All Photos(1)

Documents

15490

Sigma-Aldrich

Boc-Pro-OH

≥99.0% (T)

Synonym(s):

N-(tert-Butoxycarbonyl)-L-proline, Boc-L-proline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17NO4
CAS Number:
Molecular Weight:
215.25
Beilstein:
15828
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (T)

form

solid

optical activity

[α]20/D −61±2°, c = 2% in acetic acid

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

133-135 °C (lit.)

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N1CCC[C@H]1C(O)=O

InChI

1S/C10H17NO4/c1-10(2,3)15-9(14)11-6-4-5-7(11)8(12)13/h7H,4-6H2,1-3H3,(H,12,13)/t7-/m0/s1

InChI key

ZQEBQGAAWMOMAI-ZETCQYMHSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manos Gkikas et al.
Biomacromolecules, 16(11), 3686-3693 (2015-10-16)
Novel poly(L-lysine)-block-poly(L-proline) (PLL-b-PLP)-based materials with all PLP helical conformers, i.e., PLP II and the rare PLP I are here reported. Electrostatic supramolecular complexation of the adjacent cationic PLL with anionic molecules bearing DNA analogue H-bonding functionalities, such as deoxyguanosine monophosphate
Griet Van Zeebroeck et al.
Nature chemical biology, 5(1), 45-52 (2008-12-09)
Transporter-related nutrient sensors, called transceptors, mediate nutrient activation of signaling pathways through the plasma membrane. The mechanism of action of transporting and nontransporting transceptors is unknown. We have screened 319 amino acid analogs to identify compounds that act on Gap1

Articles

Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.

Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.

Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.

Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service