Skip to Content
Merck
All Photos(3)

Key Documents

151378

Sigma-Aldrich

Bis(4-chlorophenyl) sulfone

98%

Synonym(s):

4,4′-Dichlorodiphenyl sulfone, 4-Chlorophenyl sulfone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(ClC6H4)2SO2
CAS Number:
Molecular Weight:
287.16
Beilstein:
2052955
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

bp

250 °C/10 mmHg (lit.)

mp

143-146 °C (lit.)

functional group

chloro
sulfone

SMILES string

Clc1ccc(cc1)S(=O)(=O)c2ccc(Cl)cc2

InChI

1S/C12H8Cl2O2S/c13-9-1-5-11(6-2-9)17(15,16)12-7-3-10(14)4-8-12/h1-8H

InChI key

GPAPPPVRLPGFEQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Bis(4-chlorophenyl) sulfone was used to study the time trends of bis(4-chlorophenyl) sulfone (BCPS). It was also used in thermoplastic production.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

R S Chhabra et al.
Toxicological sciences : an official journal of the Society of Toxicology, 60(1), 28-37 (2001-02-27)
p,p'-Dichlorodiphenyl sulfone (DDS) is used as a starting material in the production of polysulfones and polyethersufones, a family of thermoplastics. DDS was studied because of its high production volume and use. In toxicology studies, 10 Fischer 344 rats and 10
R Poon et al.
Journal of toxicology and environmental health. Part A, 57(3), 185-198 (1999-06-22)
The short-term oral toxicity of a recently identified environmental pollutant, bis(4-chlorophenyl) sulfone (BCPS), was studied. Groups of male Sprague-Dawley rats (n = 6) were administered BCPS via the diet at 0 (control), 10, 100, or 1000 ppm for 4 wk.
A Olsson et al.
The Science of the total environment, 239(1-3), 19-30 (1999-11-26)
The environmental contamination situation in coastal areas of the former Soviet Union has up to now been more or less unknown. This study on perch (Perca fluviatilis), collected at three locations along the coast of Latvia during 1994 and 1995
Hrönn Jörundsdóttir et al.
Environmental pollution (Barking, Essex : 1987), 141(2), 226-237 (2005-10-18)
The dynamics of organohalogen contaminants and their metabolites are best studied over time by analysis of biota at high trophic levels. In this study, time trends, 1971-2001, of bis(4-chlorophenyl) sulfone (BCPS) and of methylsulfonyl-substituted metabolites of PCBs and 4,4'-DDE, were
[Establishment of the maximum permissible concentration of 4,4-dichlorophenylsulfone in reservoir waters].
G T Pis'ko et al.
Gigiena i sanitariia, (2)(2), 84-85 (1982-02-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service