Skip to Content
Merck
All Photos(1)

Key Documents

130273

Sigma-Aldrich

8-Nitroquinoline

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6N2O2
CAS Number:
Molecular Weight:
174.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

89-91 °C (lit.)

functional group

nitro

SMILES string

[O-][N+](=O)c1cccc2cccnc12

InChI

1S/C9H6N2O2/c12-11(13)8-5-1-3-7-4-2-6-10-9(7)8/h1-6H

InChI key

OQHHSGRZCKGLCY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

8-Nitroquinoline was used to prepare furazano [3,4-h] quinoline. It was also used to synthesize corresponding 2-substituted phenoxy-6-methoxy-8-aminoquinoline.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Chen et al.
Yao xue xue bao = Acta pharmaceutica Sinica, 27(6), 418-422 (1992-01-01)
2-Substituted phenoxy-, 4-methyl-6-methoxy-8-aminoquinolines and 7-methoxy-5-aminoquinoxaline were condensed with 1-phthalimido-bromo-alkane to yield 2-substituted phenoxy-, 4-methyl-6-methoxy-8-(1-phthalimidoalkyl)-aminoquinolines (compounds 7-10 and 15-20) and 7-methoxy-5-(1-phthalimidoalkyl)aminoquinoxalines (28-30) which were subsequently reacted with hydrazine hydrate to give 2-substituted phenoxy-, 4-methyl-6-methoxy-8-(1-aminoalkyl)-aminoquinolines (11-14 and 22-27) and 7-methoxy-5-(1-aminoalkyl) aminoquinoxalines (31-33)
M Hasegawa et al.
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 121(6), 379-393 (2001-07-04)
A reaction from various kinds of nitroquinoline with hydroxylamine in potassium hydroxide alkalinity produced a novel product, furazanoquinoline, besides the known amino derivatives. The products obtained were furazano [3,4-f] quinoline (5) from 5-nitroquinoline (1) and 6-nitroquinoline (6), and furazano [3,4-h]

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service