Recommended Products
Assay
98%
bp
100 °C/1.4 mmHg (lit.)
mp
47-50 °C (lit.)
SMILES string
Clc1cnc2ccccc2n1
InChI
1S/C8H5ClN2/c9-8-5-10-6-3-1-2-4-7(6)11-8/h1-5H
InChI key
BYHVGQHIAFURIL-UHFFFAOYSA-N
Application
2-Chloroquinoxaline (QCI) was used to study the effect of solvent on hydrolysis of QCI in aqueous–organic solvent mixtures with acetonitrile and dimethylesulphoxide. It was used in the synthesis of 2-(3-butynyl-2-methyl-2-ol) quinoxaline.It was used as reagent in the synthesis of chloroquinoxaline sulfamide.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Kinetics of the Reaction of 2-Chloro-quinoxaline with Hydroxide Ion in ACN-H2O and DMSO-H2O Binary Solvent Mixtures.
Journal of Solution Chemistry, 40(7), 1258-1270 (2011)
Inorganic chemistry, 50(20), 9804-9815 (2011-09-08)
A model system for the molybdenum cofactor has been developed that illustrates the noninnocent behavior of an N-heterocycle appended to a dithiolene chelate on molybdenum. The pyranopterin of the molybdenum cofactor is modeled by a quinoxalyldithiolene ligand (S(2)BMOQO) formed from
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service