Skip to Content
Merck
All Photos(1)

Key Documents

103772

Sigma-Aldrich

2,3,4,5,6-Pentafluorobenzoyl chloride

99%

Synonym(s):

Pentafluorobenzoic acid chloride, Perfluorobenzoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6F5COCl
CAS Number:
Molecular Weight:
230.52
Beilstein:
2054397
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.453 (lit.)

bp

158-159 °C (lit.)

density

1.601 g/mL at 25 °C (lit.)

functional group

acyl chloride
fluoro

SMILES string

Fc1c(F)c(F)c(c(F)c1F)C(Cl)=O

InChI

1S/C7ClF5O/c8-7(14)1-2(9)4(11)6(13)5(12)3(1)10

InChI key

MYHOHFDYWMPGJY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,3,4,5,6-Pentafluorobenzoyl chloride was used for derivatization of 5-androstenediol, testosterone, estradiol , dihydrotestosterone and estrone in a study to determine their serum levels. 2,3,4,5,6-Pentafluorobenzoyl chloride was used to derivatize dimethylamine(DMA) in study to develop a rapid and accurate GC–MS method for determination of DMA in human urine. 2,3,4,5,6-Pentafluorobenzoyl chloride was used in a study to detect the presence of Enterobacter cloacae in cultures of Leuconostoc mesenteroide by gas chromatography/mass spectrometry.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olga Begou et al.
Journal of pharmaceutical analysis, 10(1), 49-59 (2020-03-04)
Acetazolamide (molecular mass (MM), 222) belongs to the class of sulfonamides (R-SO2-NH2) and is one of the strongest pharmacological inhibitors of carbonic anhydrase activity. Acetazolamide is excreted unchanged in the urine. Here, we report on the development, validation and biomedical
Frank Giton et al.
European journal of endocrinology, 165(6), 917-924 (2011-09-29)
Differences in circulating steroid hormone levels have been hypothesized to explain ethnic differences in steroid-related diseases. The aim of this study was to determine the serum levels of a wide panel of steroid hormones, both androgens and estrogens, in healthy
I Elmroth et al.
Biotechnology and bioengineering, 35(8), 787-792 (1990-04-05)
Gas chromatography/mass spectrometry was used to detect the presence of Enterobacter cloacae in cultures of Leuconostoc mesenteroides, an organism used in an industrial process for production of dextrane. The penta-fluorobenzoyl-methyl ester derivative of 3-hydroxy-myristic acid, a characteristic compound of gram-negative
Hannah Colldén et al.
American journal of physiology. Endocrinology and metabolism, 317(6), E1182-E1192 (2019-11-07)
Androgens exert important effects both in androgen-responsive tissues and in the intestinal tract. To determine the impact of the gut microbiota (GM) on intestinal androgen metabolism, we measured unconjugated (free) and glucuronidated androgen levels in intestinal contents from the small
Dimitrios Tsikas et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 851(1-2), 229-239 (2006-10-03)
Dimethylamine [DMA, (CH(3))(2)NH)] is abundantly present in human urine. Main sources of urinary DMA have been reported to include trimethylamine N-oxide, a common food component, and asymmetric dimethylarginine (ADMA), an endogenous inhibitor of nitric oxide (NO) synthesis. ADMA is excreted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service