Skip to Content
Merck
All Photos(1)

Documents

126721

Sigma-Aldrich

Hydrazobenzene

Synonym(s):

N,N′-Diphenylhydrazine, N,N′-Bianiline, NSC 3510

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHNHC6H5
CAS Number:
Molecular Weight:
184.24
Beilstein:
639793
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

mp

123-126 °C (lit.)

SMILES string

N(Nc1ccccc1)c2ccccc2

InChI

1S/C12H12N2/c1-3-7-11(8-4-1)13-14-12-9-5-2-6-10-12/h1-10,13-14H

InChI key

YBQZXXMEJHZYMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Hydrazobenzene has been prepared by the reduction of azobenzene by SmI(2) in THF.
Hydrazobenzene is an intermediate in the hydrogenation of azoxybenzene to aniline.

Application

Reactant involved in:
  • Insertion reactions with organometallic tantalum complexes
  • Reduction reactions catalyzed by titanium(III) trichloride yielding amines
  • Studying the mechanism of hydrazobenzene rearrangement
  • Reaction with N-heterocyclic stable silylene
  • Synthesis of dimanganese amide hydrazide cluster complexes
  • Iron-mediated hydrazine reductions yielding iron arylimide cubanes

Other Notes

Contains varying amounts of azobenzene

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chintada Nageswara Rao et al.
The Journal of organic chemistry, 76(22), 9438-9443 (2011-10-19)
The reduction of azobenzene by SmI(2) in THF to give hydrazobenzene was investigated. The kinetics are first order in the substrate and first order in SmI(2). The kinetic order in MeOH is ca. 0.56, and in TFE it is ca.
Darol E Dodd et al.
International journal of toxicology, 31(6), 564-571 (2012-11-09)
Male F344 rats were exposed to hydrazobenzene (HZB) by dietary feed at concentrations of 0, 5, 20, 80, 200, or 300 ppm for 5 days, 2 weeks, 4 weeks, or 13 weeks duration. End points evaluated included clinical observations, body
H Fabre et al.
Journal of pharmaceutical sciences, 73(12), 1706-1709 (1984-12-01)
A high-performance liquid chromatographic method was developed for the simultaneous determination of azobenzene, hydrazobenzene, and four other decomposition products in phenylbutazone injectable formulations. Separation was achieved on a C18 column, with 0.1 M Tris-citrate buffer (pH 5.25) and acetonitrile (52:48)
Hydrazobenzene.
Report on carcinogens : carcinogen profiles, 10, 139-140 (2004-08-26)
F Matsui et al.
Journal of pharmaceutical sciences, 72(10), 1223-1224 (1983-10-01)
A high-performance liquid chromatographic method has been developed for the simultaneous determination of azobenzene and hydrazobenzene in phenylbutazone and sulfinpyrazone raw materials and formulations. The drug raw material or formulation is shaken with 1N NaOH and n-hexane and centrifuged. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service